Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: c9orf72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016262.3 | NP_001016262.2 | 5 - 6 | 534 | 65 | |
XM_012962136.1 | XP_012817590.1 | 5 - 6 | 1584 | 65 | ||
XM_012962140.1 | XP_012817594.1 | 5 - 6 | 1059 | 65 | ||
GenBank mRNA | BC135150.1 | - | 5 - 6 | 534 | 65 | |
CR761696.2 | - | 4 - 5 | 425 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |