Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nr2c2ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016306.3 | NP_001016306.1 | 1 - 2 | 132 | 66 | |
GenBank mRNA | BC129029.1 | - | 1 - 2 | 132 | 66 | |
CR761499.2 | - | 1 - 2 | 182 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |