Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nsa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017326.3 | NP_001017326.1 | 3 - 4 | 480 | 95 | |
GenBank mRNA | BC135791.1 | - | 3 - 4 | 480 | 95 | |
CR848589.2 | - | 2 - 3 | 351 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |