Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: egr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097361.1 | NP_001090830.1 | 1 - 2 | 452 | 70 | |
GenBank mRNA | BC135806.1 | - | 1 - 2 | 452 | 70 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |