Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sox10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100221.1 | NP_001093691.1 | 2 - 3 | 507 | 105 | |
XM_012960405.2 | XP_012815859.1 | 2 - 3 | 535 | 105 | ||
GenBank mRNA | BC136047.1 | - | 2 - 3 | 507 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |