Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: csdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100248.1 | NP_001093718.1 | 2 - 3 | 351 | 85 | |
XM_012960406.2 | XP_012815860.1 | 2 - 3 | 345 | 85 | ||
XM_012960407.2 | XP_012815861.1 | 2 - 3 | 341 | 85 | ||
GenBank mRNA | BC135981.1 | - | 2 - 3 | 351 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |