Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: tmeff1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100268.3 | NP_001093738.2 | 9 - 10 | 1552 | 81 | |
XM_012964170.2 | XP_012819624.1 | 8 - 9 | 1536 | 81 | ||
GenBank mRNA | BC135422.2 | - | 9 - 10 | 1386 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |