Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmem245 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012953820.2 | XP_012809274.1 | 8 - 9 | 2878 | 105 | |
XM_018089411.1 | XP_017944900.1 | 7 - 8 | 2853 | 105 | ||
GenBank mRNA | BC135929.1 | - | 7 - 8 | 1491 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |