Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: olfml2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102975.1 | NP_001096445.1 | 4 - 5 | 744 | 60 | |
GenBank mRNA | BC136146.1 | - | 4 - 5 | 744 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |