Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fabp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123411.1 | NP_001116883.1 | 2 - 3 | 285 | 73 | |
GenBank mRNA | BC161795.1 | - | 2 - 3 | 285 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |