Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ccdc51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127071.1 | NP_001120543.1 | 3 - 4 | 487 | 69 | |
XM_012960633.2 | XP_012816087.1 | 2 - 3 | 698 | 69 | ||
XM_012960634.2 | XP_012816088.1 | 4 - 5 | 1266 | 69 | ||
GenBank mRNA | BC161470.1 | - | 3 - 4 | 487 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |