Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kif19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012959642.2 | XP_012815096.1 | 7 - 8 | 1284 | 101 | |
GenBank mRNA | BC161478.1 | - | 973 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |