Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prxl2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097222.2 | NP_001090691.1 | 5 - 6 | 734 | 60 | |
GenBank mRNA | BC127374.1 | - | 5 - 6 | 633 | 60 | |
CX802408.1 | - | 5 - 6 | 734 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |