Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnajc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126759.1 | NP_001120231.1 | 6 - 7 | 496 | 99 | |
GenBank mRNA | BC160408.1 | - | 6 - 7 | 496 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |