Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fhl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126761.1 | NP_001120233.1 | 3 - 4 | 691 | 67 | |
XM_012956473.2 | XP_012811927.1 | 3 - 4 | 457 | 67 | ||
GenBank mRNA | BC160411.1 | - | 3 - 4 | 691 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |