Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: naa40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126933.1 | NP_001120405.1 | 7 - 8 | 668 | 72 | |
XM_012953533.2 | XP_012808987.1 | 7 - 8 | 664 | 72 | ||
XM_012953535.2 | XP_012808989.1 | 7 - 8 | 751 | 72 | ||
GenBank mRNA | BC161120.1 | - | 7 - 8 | 668 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |