Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: jpt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142023.1 | NP_001135495.1 | 4 - 5 | 472 | 107 | |
XM_012970172.1 | XP_012825626.1 | 4 - 5 | 581 | 107 | ||
GenBank mRNA | BC167895.1 | - | 4 - 5 | 472 | 107 | |
CU075616.1 | - | 4 - 5 | 451 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |