Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptp4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008022.1 | NP_001008023.1 | 5 | 1149 | 82 | |
XM_012962916.2 | XP_012818370.1 | 6 | 1704 | 82 | ||
XM_012962917.2 | XP_012818371.1 | 6 | 1720 | 82 | ||
XM_018093854.1 | XP_017949343.1 | 6 | 2240 | 82 | ||
GenBank mRNA | BC080902.1 | - | 5 | 1149 | 82 | |
CR855471.2 | - | 5 | 1008 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Both primers and probe map within a single exon |