Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psmb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203820.1 | NP_989151.1 | 250 | 75 | ||
GenBank mRNA | BC061603.1 | - | 250 | 75 | ||
CR761701.2 | - | 235 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis |