Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: saf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111963.1 | NP_001105433.1 | 554 | 65 | ||
GenBank mRNA | X58573.1 | - | 554 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Zea mays |