Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: asn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111997.1 | NP_001105467.1 | 550 | 66 | ||
GenBank mRNA | BT035878.1 | - | 647 | 66 | ||
FM211451.1 | - | 550 | 66 | |||
FM212633.1 | - | 553 | 66 | |||
X82849.1 | - | 550 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Zea mays |