Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ns1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111690.1 | NP_001105160.1 | 612 | 76 | ||
GenBank mRNA | AJ536578.1 | - | 612 | 76 | ||
BT066120.1 | - | 610 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Zea mays |