Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: mlo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112057.1 | NP_001105527.1 | 653 | 81 | ||
GenBank mRNA | AY029314.1 | - | 653 | 81 | ||
AY108219.1 | - | 637 | 81 |