Bacillus cereus and Pseudomonas aeruginosa.
Bacillus cereus and Pseudomonas aeruginosa stained with the LIVE BacLight™ Bacterial Gram Stain Kit (Cat. No. L7005). Gram-positive B. cereus cells fluoresce orange, whereas gram-negative P. aeruginosa cells fluoresce green.
<i>Bacillus cereus</i> and <i>Pseudomonas aeruginosa</i> stained with the LIVE <i>Bac</i>Light™ Bacterial Gram Stain Kit (Cat. No. L7005). Gram-positive <i>B. cereus</i> cells fluoresce orange, whereas gram-negative <i>P. aeruginosa</i> cells fluoresce green.
Related Products
Catalog # Name Size Price (USD) Qty
L7005 LIVE BacLight™ Bacterial Gram Stain Kit, for microscopy & quantitative assays Each
607.65
Online Exclusive
626.00
Save 18.35 (3%)
Related Images