Herasafe 2030i BSC
Thermo Scientific Herasafe 2030i Biological Safety Cabinets (BSC) are designed to maximize sample protection and user safety with exceptional containment measures, while at the same time, delivering excellent user comfort and convenience. Breakthrough innovations and unprecedented features make the Herasafe 2030i BSC our most technologically advanced and user-friendly Class II cabinet available.
Premium performance with innovative features
Motorized front window and SmartClean Plus design
Entire front sash is motorized to either open automatically to the right aperture or to completely lift up, allowing for easy access to interior of BSC. Routine maintenance, cleaning and set up tasks are simplified.
SmartClean Plus provides easy cleaning and access for large instruments
Touchscreen control and connectivity
Digital Airflow Verification (DAVe) alerts, system self-checks and performance status are provided on the full-color touchscreen. With enabled connectivity (available in select countries) the remote app provides all data you need.
Intuitive touchscreen of Herasafe 2030i biological safety cabinets
SmartFlow Plus protection
Autonomous safety features maintain proper air flow and monitor critical conditions in real time without user interaction.
SmartFlow and SmartFlow Plus technologies for advanced airflow in biosafety cabinets
Features
Herasafe 2030i BSC Feature Overview
  1. Touchscreen and connectivity* – Built-in WiFi system is ready for cloud-based capabilities like remote equipment monitoring, secure data storage, collaboration tools and more.
  2. Glass side walls and LED lighting – Enjoy a brighter workspace environment with better visibility.
  3. SmartClean Plus technology – Easily access the interior of the BSC with the motorized front sash which automatically stops at the set working height. The sash can be further opened electronically or the whole front panel hinges open for thorough cleaning access.
  4. Dual DC motors – Powering the SmartFlow Plus airflow compensation system while delivering better energy efficiency and quieter operation. Standby mode furthers energy savings and system longevity.
  5. Self-adjusting germicidal UV light intensity - Our unique cross beam UV system has exclusive self-adjusting exposure capability to enable reproducible germicidal effectiveness with every use.
  6. SmartPort convenience - Easily pass tubing and wiring through sidewall to accommodate many applications needs.
  7. Customizable worktops – Choose from work surface designs that best suit your application and work style. Standard equipment includes 2 or 4 duplex receptacles and 2 service valves.
Cell Therapy Systems series
For cell and gene therapy applications, we offer the Herasafe 2030i Biological Safety Cabinet CTS (Cell Therapy Systems) Series which consists of the product, a comprehensive documentation package including certifications and protocols, as well as compliance services.
Connectivity
The Herasafe 2030i biosafety cabinets have embedded capability to provide cloud-based connectivity*. This enables customers to check the BSC status in real time and be alerted to important issues via alarms and alerts.
Reasons to connect:
Check BSC status remotely
Analyze usage data
Receive reminders
Connectivity flyer
This flyer explains the different functionalities available with the Thermo Fisher Connect app available for Herasafe and Maxisafe 2030i BSCs. Read about where to find instrument information, energy consumption, the events log and more.
On-demand webinar
“How connected biological safety cabinets can support your lab - best practices today and future trends."
This webinar covers three reasons to connect your biological safety cabinet and current as well as future capabilities of cloud-based connectivity in BSCs.
On-demand webinar
* Availability of connectivity-enabled models depends on region. Please contact your Thermo Fisher Scientific sales representative to learn more. Thermo Fisher Connect cloud services are currently available in the following countries: Austria, Belgium, Bulgaria, Canada, Croatia, Cyprus, Czech Republic, Denmark, Estonia, Finland, France, Germany, Greece, Hungary, Iceland, Ireland, Italy, Latvia, Liechtenstein, Lithuania, Luxembourg, Malta, Netherlands, Norway, Poland, Portugal, Romania, Slovakia, Slovenia, Spain, Sweden, Switzerland, United Kingdom, United States. 
Applications
Typical markets: Biotech and Pharma, GMP environments.
Typical applications: cell culture, pharmaceutical, clinical and microbiological work where the protection of the user, the sample and the environment is required.
Certifications
NSF International certifies Thermo Scientific biological safety cabinets
NSF/ANSI 49
EN 12469 and DIN 12980 are certification standards for BSCs by TÜV Nord
TÜV GS certified according to EN 12469
LNE certifies certain Thermo Scientific biological safety cabinet models with the NF mark
LNE (catalog no. 51901254)
3D virtual tour of Herasafe 2030i BSC
360-degree, spin, zoom, rotate, discover and interact with our virtual Herasafe 2030i BSC.
Resources
Video
Video Player is loading.
Current Time 0:00
Duration 2:30
Loaded: 0%
Stream Type LIVE
Remaining Time 2:30
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Webinar
    Ordering information
    Size
    Certification
    Catalog No.
    4 ft.
    NSF
    51032334
    6 ft.
    NSF
    51032335
    0.9 m
    EN12469
    51032328
    1.2 m
    EN12469, LNE
    51032329
    1.5 m EN12469 51032330
    1.8 m EN12469 51032331