Bringing cryo-electron microscopy to you
Whether you’re an experienced microscopist, or just starting out, the Thermo Scientific Tundra Cryo-Transmission Electron Microscope (Cryo-TEM) is a dedicated structural analysis solution designed to bring cryo-electron microscopy (cryo-EM) to your laboratory at an affordable price point. Simplified data collection and rapid sample loading/unloading make the Tundra Cryo-TEM a powerful tool for answering your most challenging research questions. With the Tundra Cryo-TEM, you can obtain structures at biologically relevant resolutions in-house or reliably prepare high-quality samples for further analysis on Thermo Scientific Krios or Glacios Cryo-TEMs.
Simplified cryo-EM
The Tundra Cryo-TEM allows you to perform structural analyses on challenging proteins and macromolecules with unprecedented ease of use.
Tundra Cryo-TEM sample preparation and optimization
Cryo-EM structure determination at biologically relevant resolutions
The Tundra Cryo-TEM is a dedicated structure determination solution, operating at 100 kV, the Tundra Cryo-TEM can generate high-contrast biological images down to 2.1 Å.
Protein structures determined with the Tundra Cryo-TEM
Tundra Cryo-TEM resources
Learn more from scientists who are using the Tundra Cryo-TEM in their research and take a closer look with our on-demand product tour. 
Loading grid into the Tundra Cryo-TEM
Advanced cryo-TEM software with streamlined workflows
The Tunda Cryo-TEM comes with AI-enabled software to make it easier than ever to run and acquire high-resolution data.
Smart EPU Cryo-TEM acquisition software selecting grid squares.
Cryo-TEM service and support
To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and workflow validation.
Tundra Cryo-TEM sample loading station
Electron Microscopy Funding Support Center
A wide range of funding proposals and grants were considered when designing the Tundra Cryo-TEM to keep its price within the reach of most instrumentation grants.
Icon for Cryo-EM Funding Support center
For Research Use Only. Not for use in diagnostic procedures.