1508251251223
Which TaqMan miRNA Assay is right for you?
Choosing the best TaqMan miRNA Assay product to suit your needs depends on different factors, including the instrument available to you and the number of samples and targets being interrogated.  Consider the convenience of predesigned and pre-plated assays to save time in the lab.
There are now even more choices and formats for detection of mature, biologically active miRNA, designed to support a growing range of miRNA applications. TaqMan miRNA Assays have been configured using long-standing bioinformatics expertise in ultra-specific primer and probe design.  If you are studying a novel target or a non-model organism then you can use the Custom TaqMan Assay Design Tool to design and order a Custom TaqMan Gene Expression Assay to detect any gene from any organism.  
TaqMan AdvancedmiRNA assays employ a universal RT step for a streamlined workflow, and a universal miR-Amp step to enable highly sensitive detection by real-time PCR
TaqMan MicroRNA assays employ a novel target-specific stem–loop primer (miRNA-specific RT) during cDNA synthesis to produce a template for real-time PCR
Flexible formats
Choose from a variety of formats and configurations to meet all your different research needs. Whether you are looking at a single target or thousands of samples, there is a TaqMan Assay format for you.
single-tubes-150
Single tubes
  • Low entry price
  • Flexible
  • Run on any real-time PCR instrument
384-well-microfluidic-cards-150
384-well microfluidic cards
  • Low cost per reaction
  • Optimal for medium to large projects
  • Run on Applied Biosystems™ QuantStudio™ 7 Flex & 12K Flex, ViiA™ 7, and 7900HT Fast real-time PCR systems
96-or-384-well-plates-150
96-well plates
  • Optimal for small to medium projects
  • Balances flexibility with streamlined reaction setup
  • Run on any 96-well real-time PCR instrument
openarray-plates-150
OpenArray plates
  • Lowest cost for large projects
  • Ultimate throughput
  • Run on QuantStudio 12K Flex real-time PCR system
If you are planning on duplexing or multiplexing, make sure that the reporter dyes you choose are compatible with your qPCR instrument model. Each instrument must have an excitation source to excite the fluorescent dye and a detector to detect the fluorescent emissions. The excitation and emission filters support different types of dyes, and the number of these filters can vary depending upon the qPCR instrument model chosen.
For miRNA studies, selection of a valid endogenous control is critical for normalization and corrects for inter-sample variation.  We offer endogenous controls in all formats to help you determine the best control gene for your study.
TaqMan miRNA Assay selection tool
Find the right TaqMan miRNA Assay for your needs.
instrument
  • 7300 Real-Time PCR System
  • 7500 Real-Time PCR System
  • 7500 Fast Real-Time PCR System
  • 7900HT Real-Time PCR System
  • QuantStudio 3 System
  • QuantStudio 5 System
  • QuantStudio 6 Flex System
  • QuantStudio 7 Flex System
  • QuantStudio 12K Flex System
  • StepOnePlus System
  • ViiA7 System
  • Analytik Jena qTOWER
  • Bio-Rad CFX384
  • Bio-Rad CFX96
  • Bio-Rad iQ5
  • Bio-Rad MiniOpticon
  • Bio-Rad/MJ Chromo4
  • Bio-Rad/MJ Opticon
  • Bio-Rad/MJ Opticon 2
  • Eppendorf Mastercycler ep realplex
  • Qiagen/Corbett Rotor-Gene 6000
  • Qiagen/Corbett Rotor-Gene Q
  • Qiagen/Corbett Rotor-Gene 3000
  • Roche LightCycler 480
  • Roche LightCycler Nano
  • Agilent/Stratagene MX3000P
  • Agilent/Stratagene MX3005P
  • Agilent/Stratagene MX4000
  • TaKaRa T800
block
  • 96-well std
  • 96-well fast
  • 384-well
  • N/A
  • 96-well
  • TaqMan Array
  • OpenArray
format
  • Single tube
  • std
  • 96-well Fast Plate
  • fast
  • 96-well Standard Plate
  • 384 well microfluidic card
  • 3072 Through holes
panel category
  • TaqMan Advanced miRNA Assays
  • TaqMan miRNA Assays
  • Custom TaqMan Small RNA Assays
  • Custom TaqMan Advanced Small RNA Assays
  • TaqMan™ Advanced miRNA Human Endogenous Controls 96-well Plate, standard
  • TaqMan™ Advanced miRNA Human Endogenous Controls 96-well Plate, fast
  • TaqMan™ Advanced miRNA Human Fixed Content 96-well Plate, standard
  • TaqMan™ Advanced miRNA Human Fixed Content 96-well Plate, fast
  • Pre-Configured Content TaqMan Advanced miRNA Cards
  • Custom Content TaqMan Advanced miRNA Cards
  • TaqMan™ OpenArray™ Human Advanced MicroRNA Panel, QuantStudio™ 12K Flex
  • TaqMan™ OpenArray™ Rodent Advanced MicroRNA Panel, QuantStudio™ 12K Flex
InstrumentBlockFormatPredesigned or Custom AssaysRT ChemistryMinimum Order QtyContent TypeProduct
7300 Real-Time PCR System96-well stdSingle tubePredesignedUniversal RT1allTaqMan Advanced miRNA Assays
7500 Real-Time PCR System96-well stdSingle tubePredesignedUniversal RT1allTaqMan Advanced miRNA Assays
7500 Fast Real-Time PCR System96-well fastSingle tubePredesignedUniversal RT1allTaqMan Advanced miRNA Assays
7900HT Real-Time PCR System96-well stdSingle tubePredesignedUniversal RT1allTaqMan Advanced miRNA Assays
7900HT Real-Time PCR System96-well fastSingle tubePredesignedUniversal RT1allTaqMan Advanced miRNA Assays
7900HT Real-Time PCR System384-wellSingle tubePredesignedUniversal RT1allTaqMan Advanced miRNA Assays
QuantStudio 3 System96-well stdSingle tubePredesignedUniversal RT1allTaqMan Advanced miRNA Assays
QuantStudio 3 System96-well fastSingle tubePredesignedUniversal RT1allTaqMan Advanced miRNA Assays
QuantStudio 5 System96-well stdSingle tubePredesignedUniversal RT1allTaqMan Advanced miRNA Assays
QuantStudio 5 System96-well fastSingle tubePredesignedUniversal RT1allTaqMan Advanced miRNA Assays
QuantStudio 5 System384-wellSingle tubePredesignedUniversal RT1allTaqMan Advanced miRNA Assays
QuantStudio 6 Flex System96-well stdSingle tubePredesignedUniversal RT1allTaqMan Advanced miRNA Assays
QuantStudio 6 Flex System96-well fastSingle tubePredesignedUniversal RT1allTaqMan Advanced miRNA Assays
QuantStudio 6 Flex System384-wellSingle tubePredesignedUniversal RT1allTaqMan Advanced miRNA Assays
QuantStudio 7 Flex System96-well stdSingle tubePredesignedUniversal RT1allTaqMan Advanced miRNA Assays
QuantStudio 7 Flex System96-well fastSingle tubePredesignedUniversal RT1allTaqMan Advanced miRNA Assays
QuantStudio 7 Flex System384-wellSingle tubePredesignedUniversal RT1allTaqMan Advanced miRNA Assays
QuantStudio 12K Flex System96-well stdSingle tubePredesignedUniversal RT1allTaqMan Advanced miRNA Assays
QuantStudio 12K Flex System96-well fastSingle tubePredesignedUniversal RT1allTaqMan Advanced miRNA Assays
QuantStudio 12K Flex System384-wellSingle tubePredesignedUniversal RT1allTaqMan Advanced miRNA Assays
StepOnePlus System96-well fastSingle tubePredesignedUniversal RT1allTaqMan Advanced miRNA Assays
ViiA7 System96-well stdSingle tubePredesignedUniversal RT1allTaqMan Advanced miRNA Assays
ViiA7 System96-well fastSingle tubePredesignedUniversal RT1allTaqMan Advanced miRNA Assays
ViiA7 System384-wellSingle tubePredesignedUniversal RT1allTaqMan Advanced miRNA Assays
Analytik Jena qTOWERN/ASingle tubePredesignedUniversal RT1allTaqMan Advanced miRNA Assays
Bio-Rad CFX384384-wellSingle tubePredesignedUniversal RT1allTaqMan Advanced miRNA Assays
Bio-Rad CFX9696-wellSingle tubePredesignedUniversal RT1allTaqMan Advanced miRNA Assays
Bio-Rad iQ5N/ASingle tubePredesignedUniversal RT1allTaqMan Advanced miRNA Assays
Bio-Rad MiniOpticonN/ASingle tubePredesignedUniversal RT1allTaqMan Advanced miRNA Assays
Bio-Rad/MJ Chromo4N/ASingle tubePredesignedUniversal RT1allTaqMan Advanced miRNA Assays
Bio-Rad/MJ OpticonN/ASingle tubePredesignedUniversal RT1allTaqMan Advanced miRNA Assays
Bio-Rad/MJ Opticon 2N/ASingle tubePredesignedUniversal RT1allTaqMan Advanced miRNA Assays
Eppendorf Mastercycler ep realplexN/ASingle tubePredesignedUniversal RT1allTaqMan Advanced miRNA Assays
Qiagen/Corbett Rotor-Gene 6000N/ASingle tubePredesignedUniversal RT1allTaqMan Advanced miRNA Assays
Qiagen/Corbett Rotor-Gene QN/ASingle tubePredesignedUniversal RT1allTaqMan Advanced miRNA Assays
Qiagen/Corbett Rotor-Gene 3000N/ASingle tubePredesignedUniversal RT1allTaqMan Advanced miRNA Assays
Roche LightCycler 480N/ASingle tubePredesignedUniversal RT1allTaqMan Advanced miRNA Assays
Roche LightCycler NanoN/ASingle tubePredesignedUniversal RT1allTaqMan Advanced miRNA Assays
Agilent/Stratagene MX3000PN/ASingle tubePredesignedUniversal RT1allTaqMan Advanced miRNA Assays
Agilent/Stratagene MX3005PN/ASingle tubePredesignedUniversal RT1allTaqMan Advanced miRNA Assays
Agilent/Stratagene MX4000N/ASingle tubePredesignedUniversal RT1allTaqMan Advanced miRNA Assays
TaKaRa T800N/ASingle tubePredesignedUniversal RT1allTaqMan Advanced miRNA Assays
7300 Real-Time PCR System96-well stdSingle tubePredesignedUniversal RT1ControlsTaqMan Advanced miRNA Assays
7500 Real-Time PCR System96-well stdSingle tubePredesignedUniversal RT1ControlsTaqMan Advanced miRNA Assays
7500 Fast Real-Time PCR System96-well fastSingle tubePredesignedUniversal RT1ControlsTaqMan Advanced miRNA Assays
7900HT Real-Time PCR System96-well stdSingle tubePredesignedUniversal RT1ControlsTaqMan Advanced miRNA Assays
7900HT Real-Time PCR System96-well fastSingle tubePredesignedUniversal RT1ControlsTaqMan Advanced miRNA Assays
7900HT Real-Time PCR System384-wellSingle tubePredesignedUniversal RT1ControlsTaqMan Advanced miRNA Assays
QuantStudio 3 System96-well stdSingle tubePredesignedUniversal RT1ControlsTaqMan Advanced miRNA Assays
QuantStudio 3 System96-well fastSingle tubePredesignedUniversal RT1ControlsTaqMan Advanced miRNA Assays
QuantStudio 5 System96-well stdSingle tubePredesignedUniversal RT1ControlsTaqMan Advanced miRNA Assays
QuantStudio 5 System96-well fastSingle tubePredesignedUniversal RT1ControlsTaqMan Advanced miRNA Assays
QuantStudio 5 System384-wellSingle tubePredesignedUniversal RT1ControlsTaqMan Advanced miRNA Assays
QuantStudio 6 Flex System96-well stdSingle tubePredesignedUniversal RT1ControlsTaqMan Advanced miRNA Assays
QuantStudio 6 Flex System96-well fastSingle tubePredesignedUniversal RT1ControlsTaqMan Advanced miRNA Assays
QuantStudio 6 Flex System384-wellSingle tubePredesignedUniversal RT1ControlsTaqMan Advanced miRNA Assays
QuantStudio 7 Flex System96-well stdSingle tubePredesignedUniversal RT1ControlsTaqMan Advanced miRNA Assays
QuantStudio 7 Flex System96-well fastSingle tubePredesignedUniversal RT1ControlsTaqMan Advanced miRNA Assays
QuantStudio 7 Flex System384-wellSingle tubePredesignedUniversal RT1ControlsTaqMan Advanced miRNA Assays
QuantStudio 12K Flex System96-well stdSingle tubePredesignedUniversal RT1ControlsTaqMan Advanced miRNA Assays
QuantStudio 12K Flex System96-well fastSingle tubePredesignedUniversal RT1ControlsTaqMan Advanced miRNA Assays
QuantStudio 12K Flex System384-wellSingle tubePredesignedUniversal RT1ControlsTaqMan Advanced miRNA Assays
StepOnePlus System96-well fastSingle tubePredesignedUniversal RT1ControlsTaqMan Advanced miRNA Assays
ViiA7 System96-well stdSingle tubePredesignedUniversal RT1ControlsTaqMan Advanced miRNA Assays
ViiA7 System96-well fastSingle tubePredesignedUniversal RT1ControlsTaqMan Advanced miRNA Assays
ViiA7 System384-wellSingle tubePredesignedUniversal RT1ControlsTaqMan Advanced miRNA Assays
Analytik Jena qTOWERN/ASingle tubePredesignedUniversal RT1ControlsTaqMan Advanced miRNA Assays
Bio-Rad CFX384384-wellSingle tubePredesignedUniversal RT1ControlsTaqMan Advanced miRNA Assays
Bio-Rad CFX9696-wellSingle tubePredesignedUniversal RT1ControlsTaqMan Advanced miRNA Assays
Bio-Rad iQ5N/ASingle tubePredesignedUniversal RT1ControlsTaqMan Advanced miRNA Assays
Bio-Rad MiniOpticonN/ASingle tubePredesignedUniversal RT1ControlsTaqMan Advanced miRNA Assays
Bio-Rad/MJ Chromo4N/ASingle tubePredesignedUniversal RT1ControlsTaqMan Advanced miRNA Assays
Bio-Rad/MJ OpticonN/ASingle tubePredesignedUniversal RT1ControlsTaqMan Advanced miRNA Assays
Bio-Rad/MJ Opticon 2N/ASingle tubePredesignedUniversal RT1ControlsTaqMan Advanced miRNA Assays
Eppendorf Mastercycler ep realplexN/ASingle tubePredesignedUniversal RT1ControlsTaqMan Advanced miRNA Assays
Qiagen/Corbett Rotor-Gene 6000N/ASingle tubePredesignedUniversal RT1ControlsTaqMan Advanced miRNA Assays
Qiagen/Corbett Rotor-Gene QN/ASingle tubePredesignedUniversal RT1ControlsTaqMan Advanced miRNA Assays
Qiagen/Corbett Rotor-Gene 3000N/ASingle tubePredesignedUniversal RT1ControlsTaqMan Advanced miRNA Assays
Roche LightCycler 480N/ASingle tubePredesignedUniversal RT1ControlsTaqMan Advanced miRNA Assays
Roche LightCycler NanoN/ASingle tubePredesignedUniversal RT1ControlsTaqMan Advanced miRNA Assays
Agilent/Stratagene MX3000PN/ASingle tubePredesignedUniversal RT1ControlsTaqMan Advanced miRNA Assays
Agilent/Stratagene MX3005PN/ASingle tubePredesignedUniversal RT1ControlsTaqMan Advanced miRNA Assays
Agilent/Stratagene MX4000N/ASingle tubePredesignedUniversal RT1ControlsTaqMan Advanced miRNA Assays
TaKaRa T800N/ASingle tubePredesignedUniversal RT1ControlsTaqMan Advanced miRNA Assays
7300 Real-Time PCR System96-well stdSingle tubePredesignedmiRNA-specific RT1allTaqMan miRNA Assays
7500 Real-Time PCR System96-well stdSingle tubePredesignedmiRNA-specific RT1allTaqMan miRNA Assays
7500 Fast Real-Time PCR System96-well fastSingle tubePredesignedmiRNA-specific RT1allTaqMan miRNA Assays
7900HT Real-Time PCR System96-well stdSingle tubePredesignedmiRNA-specific RT1allTaqMan miRNA Assays
7900HT Real-Time PCR System96-well fastSingle tubePredesignedmiRNA-specific RT1allTaqMan miRNA Assays
7900HT Real-Time PCR System384-wellSingle tubePredesignedmiRNA-specific RT1allTaqMan miRNA Assays
QuantStudio 3 System96-well stdSingle tubePredesignedmiRNA-specific RT1allTaqMan miRNA Assays
QuantStudio 3 System96-well fastSingle tubePredesignedmiRNA-specific RT1allTaqMan miRNA Assays
QuantStudio 5 System96-well stdSingle tubePredesignedmiRNA-specific RT1allTaqMan miRNA Assays
QuantStudio 5 System96-well fastSingle tubePredesignedmiRNA-specific RT1allTaqMan miRNA Assays
QuantStudio 5 System384-wellSingle tubePredesignedmiRNA-specific RT1allTaqMan miRNA Assays
QuantStudio 6 Flex System96-well stdSingle tubePredesignedmiRNA-specific RT1allTaqMan miRNA Assays
QuantStudio 6 Flex System96-well fastSingle tubePredesignedmiRNA-specific RT1allTaqMan miRNA Assays
QuantStudio 6 Flex System384-wellSingle tubePredesignedmiRNA-specific RT1allTaqMan miRNA Assays
QuantStudio 7 Flex System96-well stdSingle tubePredesignedmiRNA-specific RT1allTaqMan miRNA Assays
QuantStudio 7 Flex System96-well fastSingle tubePredesignedmiRNA-specific RT1allTaqMan miRNA Assays
QuantStudio 7 Flex System384-wellSingle tubePredesignedmiRNA-specific RT1allTaqMan miRNA Assays
QuantStudio 12K Flex System96-well stdSingle tubePredesignedmiRNA-specific RT1allTaqMan miRNA Assays
QuantStudio 12K Flex System96-well fastSingle tubePredesignedmiRNA-specific RT1allTaqMan miRNA Assays
QuantStudio 12K Flex System384-wellSingle tubePredesignedmiRNA-specific RT1allTaqMan miRNA Assays
StepOnePlus System96-well fastSingle tubePredesignedmiRNA-specific RT1allTaqMan miRNA Assays
ViiA7 System96-well stdSingle tubePredesignedmiRNA-specific RT1allTaqMan miRNA Assays
ViiA7 System96-well fastSingle tubePredesignedmiRNA-specific RT1allTaqMan miRNA Assays
ViiA7 System384-wellSingle tubePredesignedmiRNA-specific RT1allTaqMan miRNA Assays
Analytik Jena qTOWERN/ASingle tubePredesignedmiRNA-specific RT1allTaqMan miRNA Assays
Bio-Rad CFX384384-wellSingle tubePredesignedmiRNA-specific RT1allTaqMan miRNA Assays
Bio-Rad CFX9696-wellSingle tubePredesignedmiRNA-specific RT1allTaqMan miRNA Assays
Bio-Rad iQ5N/ASingle tubePredesignedmiRNA-specific RT1allTaqMan miRNA Assays
Bio-Rad MiniOpticonN/ASingle tubePredesignedmiRNA-specific RT1allTaqMan miRNA Assays
Bio-Rad/MJ Chromo4N/ASingle tubePredesignedmiRNA-specific RT1allTaqMan miRNA Assays
Bio-Rad/MJ OpticonN/ASingle tubePredesignedmiRNA-specific RT1allTaqMan miRNA Assays
Bio-Rad/MJ Opticon 2N/ASingle tubePredesignedmiRNA-specific RT1allTaqMan miRNA Assays
Eppendorf Mastercycler ep realplexN/ASingle tubePredesignedmiRNA-specific RT1allTaqMan miRNA Assays
Qiagen/Corbett Rotor-Gene 6000N/ASingle tubePredesignedmiRNA-specific RT1allTaqMan miRNA Assays
Qiagen/Corbett Rotor-Gene QN/ASingle tubePredesignedmiRNA-specific RT1allTaqMan miRNA Assays
Qiagen/Corbett Rotor-Gene 3000N/ASingle tubePredesignedmiRNA-specific RT1allTaqMan miRNA Assays
Roche LightCycler 480N/ASingle tubePredesignedmiRNA-specific RT1allTaqMan miRNA Assays
Roche LightCycler NanoN/ASingle tubePredesignedmiRNA-specific RT1allTaqMan miRNA Assays
Agilent/Stratagene MX3000PN/ASingle tubePredesignedmiRNA-specific RT1allTaqMan miRNA Assays
Agilent/Stratagene MX3005PN/ASingle tubePredesignedmiRNA-specific RT1allTaqMan miRNA Assays
Agilent/Stratagene MX4000N/ASingle tubePredesignedmiRNA-specific RT1allTaqMan miRNA Assays
TaKaRa T800N/ASingle tubePredesignedmiRNA-specific RT1allTaqMan miRNA Assays
7300 Real-Time PCR System96-well stdSingle tubePredesignedmiRNA-specific RT1ControlsTaqMan miRNA Assays
7500 Real-Time PCR System96-well stdSingle tubePredesignedmiRNA-specific RT1ControlsTaqMan miRNA Assays
7500 Fast Real-Time PCR System96-well fastSingle tubePredesignedmiRNA-specific RT1ControlsTaqMan miRNA Assays
7900HT Real-Time PCR System96-well stdSingle tubePredesignedmiRNA-specific RT1ControlsTaqMan miRNA Assays
7900HT Real-Time PCR System96-well fastSingle tubePredesignedmiRNA-specific RT1ControlsTaqMan miRNA Assays
7900HT Real-Time PCR System384-wellSingle tubePredesignedmiRNA-specific RT1ControlsTaqMan miRNA Assays
QuantStudio 3 System96-well stdSingle tubePredesignedmiRNA-specific RT1ControlsTaqMan miRNA Assays
QuantStudio 3 System96-well fastSingle tubePredesignedmiRNA-specific RT1ControlsTaqMan miRNA Assays
QuantStudio 5 System96-well stdSingle tubePredesignedmiRNA-specific RT1ControlsTaqMan miRNA Assays
QuantStudio 5 System96-well fastSingle tubePredesignedmiRNA-specific RT1ControlsTaqMan miRNA Assays
QuantStudio 5 System384-wellSingle tubePredesignedmiRNA-specific RT1ControlsTaqMan miRNA Assays
QuantStudio 6 Flex System96-well stdSingle tubePredesignedmiRNA-specific RT1ControlsTaqMan miRNA Assays
QuantStudio 6 Flex System96-well fastSingle tubePredesignedmiRNA-specific RT1ControlsTaqMan miRNA Assays
QuantStudio 6 Flex System384-wellSingle tubePredesignedmiRNA-specific RT1ControlsTaqMan miRNA Assays
QuantStudio 7 Flex System96-well stdSingle tubePredesignedmiRNA-specific RT1ControlsTaqMan miRNA Assays
QuantStudio 7 Flex System96-well fastSingle tubePredesignedmiRNA-specific RT1ControlsTaqMan miRNA Assays
QuantStudio 7 Flex System384-wellSingle tubePredesignedmiRNA-specific RT1ControlsTaqMan miRNA Assays
QuantStudio 12K Flex System96-well stdSingle tubePredesignedmiRNA-specific RT1ControlsTaqMan miRNA Assays
QuantStudio 12K Flex System96-well fastSingle tubePredesignedmiRNA-specific RT1ControlsTaqMan miRNA Assays
QuantStudio 12K Flex System384-wellSingle tubePredesignedmiRNA-specific RT1ControlsTaqMan miRNA Assays
StepOnePlus System96-well fastSingle tubePredesignedmiRNA-specific RT1ControlsTaqMan miRNA Assays
ViiA7 System96-well stdSingle tubePredesignedmiRNA-specific RT1ControlsTaqMan miRNA Assays
ViiA7 System96-well fastSingle tubePredesignedmiRNA-specific RT1ControlsTaqMan miRNA Assays
ViiA7 System384-wellSingle tubePredesignedmiRNA-specific RT1ControlsTaqMan miRNA Assays
Analytik Jena qTOWERN/ASingle tubePredesignedmiRNA-specific RT1ControlsTaqMan miRNA Assays
Bio-Rad CFX384384-wellSingle tubePredesignedmiRNA-specific RT1ControlsTaqMan miRNA Assays
Bio-Rad CFX9696-wellSingle tubePredesignedmiRNA-specific RT1ControlsTaqMan miRNA Assays
Bio-Rad iQ5N/ASingle tubePredesignedmiRNA-specific RT1ControlsTaqMan miRNA Assays
Bio-Rad MiniOpticonN/ASingle tubePredesignedmiRNA-specific RT1ControlsTaqMan miRNA Assays
Bio-Rad/MJ Chromo4N/ASingle tubePredesignedmiRNA-specific RT1ControlsTaqMan miRNA Assays
Bio-Rad/MJ OpticonN/ASingle tubePredesignedmiRNA-specific RT1ControlsTaqMan miRNA Assays
Bio-Rad/MJ Opticon 2N/ASingle tubePredesignedmiRNA-specific RT1ControlsTaqMan miRNA Assays
Eppendorf Mastercycler ep realplexN/ASingle tubePredesignedmiRNA-specific RT1ControlsTaqMan miRNA Assays
Qiagen/Corbett Rotor-Gene 6000N/ASingle tubePredesignedmiRNA-specific RT1ControlsTaqMan miRNA Assays
Qiagen/Corbett Rotor-Gene QN/ASingle tubePredesignedmiRNA-specific RT1ControlsTaqMan miRNA Assays
Qiagen/Corbett Rotor-Gene 3000N/ASingle tubePredesignedmiRNA-specific RT1ControlsTaqMan miRNA Assays
Roche LightCycler 480N/ASingle tubePredesignedmiRNA-specific RT1ControlsTaqMan miRNA Assays
Roche LightCycler NanoN/ASingle tubePredesignedmiRNA-specific RT1ControlsTaqMan miRNA Assays
Agilent/Stratagene MX3000PN/ASingle tubePredesignedmiRNA-specific RT1ControlsTaqMan miRNA Assays
Agilent/Stratagene MX3005PN/ASingle tubePredesignedmiRNA-specific RT1ControlsTaqMan miRNA Assays
Agilent/Stratagene MX4000N/ASingle tubePredesignedmiRNA-specific RT1ControlsTaqMan miRNA Assays
TaKaRa T800N/ASingle tubePredesignedmiRNA-specific RT1ControlsTaqMan miRNA Assays
7300 Real-Time PCR System96-well stdSingle tubeCustommiRNA-specific RT1allCustom TaqMan Small RNA Assays
7500 Real-Time PCR System96-well stdSingle tubeCustommiRNA-specific RT1allCustom TaqMan Small RNA Assays
7500 Fast Real-Time PCR System96-well fastSingle tubeCustommiRNA-specific RT1allCustom TaqMan Small RNA Assays
7900HT Real-Time PCR System96-well stdSingle tubeCustommiRNA-specific RT1allCustom TaqMan Small RNA Assays
7900HT Real-Time PCR System96-well fastSingle tubeCustommiRNA-specific RT1allCustom TaqMan Small RNA Assays
7900HT Real-Time PCR System384-wellSingle tubeCustommiRNA-specific RT1allCustom TaqMan Small RNA Assays
QuantStudio 3 System96-well stdSingle tubeCustommiRNA-specific RT1allCustom TaqMan Small RNA Assays
QuantStudio 3 System96-well fastSingle tubeCustommiRNA-specific RT1allCustom TaqMan Small RNA Assays
QuantStudio 5 System96-well stdSingle tubeCustommiRNA-specific RT1allCustom TaqMan Small RNA Assays
QuantStudio 5 System96-well fastSingle tubeCustommiRNA-specific RT1allCustom TaqMan Small RNA Assays
QuantStudio 5 System384-wellSingle tubeCustommiRNA-specific RT1allCustom TaqMan Small RNA Assays
QuantStudio 6 Flex System96-well stdSingle tubeCustommiRNA-specific RT1allCustom TaqMan Small RNA Assays
QuantStudio 6 Flex System96-well fastSingle tubeCustommiRNA-specific RT1allCustom TaqMan Small RNA Assays
QuantStudio 6 Flex System384-wellSingle tubeCustommiRNA-specific RT1allCustom TaqMan Small RNA Assays
QuantStudio 7 Flex System96-well stdSingle tubeCustommiRNA-specific RT1allCustom TaqMan Small RNA Assays
QuantStudio 7 Flex System96-well fastSingle tubeCustommiRNA-specific RT1allCustom TaqMan Small RNA Assays
QuantStudio 7 Flex System384-wellSingle tubeCustommiRNA-specific RT1allCustom TaqMan Small RNA Assays
QuantStudio 12K Flex System96-well stdSingle tubeCustommiRNA-specific RT1allCustom TaqMan Small RNA Assays
QuantStudio 12K Flex System96-well fastSingle tubeCustommiRNA-specific RT1allCustom TaqMan Small RNA Assays
QuantStudio 12K Flex System384-wellSingle tubeCustommiRNA-specific RT1allCustom TaqMan Small RNA Assays
StepOnePlus System96-well fastSingle tubeCustommiRNA-specific RT1allCustom TaqMan Small RNA Assays
ViiA7 System96-well stdSingle tubeCustommiRNA-specific RT1allCustom TaqMan Small RNA Assays
ViiA7 System96-well fastSingle tubeCustommiRNA-specific RT1allCustom TaqMan Small RNA Assays
ViiA7 System384-wellSingle tubeCustommiRNA-specific RT1allCustom TaqMan Small RNA Assays
Analytik Jena qTOWERN/ASingle tubeCustommiRNA-specific RT1allCustom TaqMan Small RNA Assays
Bio-Rad CFX384384-wellSingle tubeCustommiRNA-specific RT1allCustom TaqMan Small RNA Assays
Bio-Rad CFX9696-wellSingle tubeCustommiRNA-specific RT1allCustom TaqMan Small RNA Assays
Bio-Rad iQ5N/ASingle tubeCustommiRNA-specific RT1allCustom TaqMan Small RNA Assays
Bio-Rad MiniOpticonN/ASingle tubeCustommiRNA-specific RT1allCustom TaqMan Small RNA Assays
Bio-Rad/MJ Chromo4N/ASingle tubeCustommiRNA-specific RT1allCustom TaqMan Small RNA Assays
Bio-Rad/MJ OpticonN/ASingle tubeCustommiRNA-specific RT1allCustom TaqMan Small RNA Assays
Bio-Rad/MJ Opticon 2N/ASingle tubeCustommiRNA-specific RT1allCustom TaqMan Small RNA Assays
Eppendorf Mastercycler ep realplexN/ASingle tubeCustommiRNA-specific RT1allCustom TaqMan Small RNA Assays
Qiagen/Corbett Rotor-Gene 6000N/ASingle tubeCustommiRNA-specific RT1allCustom TaqMan Small RNA Assays
Qiagen/Corbett Rotor-Gene QN/ASingle tubeCustommiRNA-specific RT1allCustom TaqMan Small RNA Assays
Qiagen/Corbett Rotor-Gene 3000N/ASingle tubeCustommiRNA-specific RT1allCustom TaqMan Small RNA Assays
Roche LightCycler 480N/ASingle tubeCustommiRNA-specific RT1allCustom TaqMan Small RNA Assays
Roche LightCycler NanoN/ASingle tubeCustommiRNA-specific RT1allCustom TaqMan Small RNA Assays
Agilent/Stratagene MX3000PN/ASingle tubeCustommiRNA-specific RT1allCustom TaqMan Small RNA Assays
Agilent/Stratagene MX3005PN/ASingle tubeCustommiRNA-specific RT1allCustom TaqMan Small RNA Assays
Agilent/Stratagene MX4000N/ASingle tubeCustommiRNA-specific RT1allCustom TaqMan Small RNA Assays
TaKaRa T800N/ASingle tubeCustommiRNA-specific RT1allCustom TaqMan Small RNA Assays
7300 Real-Time PCR System96-well stdSingle tubeCustomUniversal RT1allCustom TaqMan Advanced Small RNA Assays
7500 Real-Time PCR System96-well stdSingle tubeCustomUniversal RT1allCustom TaqMan Advanced Small RNA Assays
7500 Fast Real-Time PCR System96-well fastSingle tubeCustomUniversal RT1allCustom TaqMan Advanced Small RNA Assays
7900HT Real-Time PCR System96-well stdSingle tubeCustomUniversal RT1allCustom TaqMan Advanced Small RNA Assays
7900HT Real-Time PCR System96-well fastSingle tubeCustomUniversal RT1allCustom TaqMan Advanced Small RNA Assays
7900HT Real-Time PCR System384-wellSingle tubeCustomUniversal RT1allCustom TaqMan Advanced Small RNA Assays
QuantStudio 3 System96-well stdSingle tubeCustomUniversal RT1allCustom TaqMan Advanced Small RNA Assays
QuantStudio 3 System96-well fastSingle tubeCustomUniversal RT1allCustom TaqMan Advanced Small RNA Assays
QuantStudio 5 System96-well stdSingle tubeCustomUniversal RT1allCustom TaqMan Advanced Small RNA Assays
QuantStudio 5 System96-well fastSingle tubeCustomUniversal RT1allCustom TaqMan Advanced Small RNA Assays
QuantStudio 5 System384-wellSingle tubeCustomUniversal RT1allCustom TaqMan Advanced Small RNA Assays
QuantStudio 6 Flex System96-well stdSingle tubeCustomUniversal RT1allCustom TaqMan Advanced Small RNA Assays
QuantStudio 6 Flex System96-well fastSingle tubeCustomUniversal RT1allCustom TaqMan Advanced Small RNA Assays
QuantStudio 6 Flex System384-wellSingle tubeCustomUniversal RT1allCustom TaqMan Advanced Small RNA Assays
QuantStudio 7 Flex System96-well stdSingle tubeCustomUniversal RT1allCustom TaqMan Advanced Small RNA Assays
QuantStudio 7 Flex System96-well fastSingle tubeCustomUniversal RT1allCustom TaqMan Advanced Small RNA Assays
QuantStudio 7 Flex System384-wellSingle tubeCustomUniversal RT1allCustom TaqMan Advanced Small RNA Assays
QuantStudio 12K Flex System96-well stdSingle tubeCustomUniversal RT1allCustom TaqMan Advanced Small RNA Assays
QuantStudio 12K Flex System96-well fastSingle tubeCustomUniversal RT1allCustom TaqMan Advanced Small RNA Assays
QuantStudio 12K Flex System384-wellSingle tubeCustomUniversal RT1allCustom TaqMan Advanced Small RNA Assays
StepOnePlus System96-well fastSingle tubeCustomUniversal RT1allCustom TaqMan Advanced Small RNA Assays
ViiA7 System96-well stdSingle tubeCustomUniversal RT1allCustom TaqMan Advanced Small RNA Assays
ViiA7 System96-well fastSingle tubeCustomUniversal RT1allCustom TaqMan Advanced Small RNA Assays
ViiA7 System384-wellSingle tubeCustomUniversal RT1allCustom TaqMan Advanced Small RNA Assays
Analytik Jena qTOWERN/ASingle tubeCustomUniversal RT1allCustom TaqMan Advanced Small RNA Assays
Bio-Rad CFX384384-wellSingle tubeCustomUniversal RT1allCustom TaqMan Advanced Small RNA Assays
Bio-Rad CFX9696-wellSingle tubeCustomUniversal RT1allCustom TaqMan Advanced Small RNA Assays
Bio-Rad iQ5N/ASingle tubeCustomUniversal RT1allCustom TaqMan Advanced Small RNA Assays
Bio-Rad MiniOpticonN/ASingle tubeCustomUniversal RT1allCustom TaqMan Advanced Small RNA Assays
Bio-Rad/MJ Chromo4N/ASingle tubeCustomUniversal RT1allCustom TaqMan Advanced Small RNA Assays
Bio-Rad/MJ OpticonN/ASingle tubeCustomUniversal RT1allCustom TaqMan Advanced Small RNA Assays
Bio-Rad/MJ Opticon 2N/ASingle tubeCustomUniversal RT1allCustom TaqMan Advanced Small RNA Assays
Eppendorf Mastercycler ep realplexN/ASingle tubeCustomUniversal RT1allCustom TaqMan Advanced Small RNA Assays
Qiagen/Corbett Rotor-Gene 6000N/ASingle tubeCustomUniversal RT1allCustom TaqMan Advanced Small RNA Assays
Qiagen/Corbett Rotor-Gene QN/ASingle tubeCustomUniversal RT1allCustom TaqMan Advanced Small RNA Assays
Qiagen/Corbett Rotor-Gene 3000N/ASingle tubeCustomUniversal RT1allCustom TaqMan Advanced Small RNA Assays
Roche LightCycler 480N/ASingle tubeCustomUniversal RT1allCustom TaqMan Advanced Small RNA Assays
Roche LightCycler NanoN/ASingle tubeCustomUniversal RT1allCustom TaqMan Advanced Small RNA Assays
Agilent/Stratagene MX3000PN/ASingle tubeCustomUniversal RT1allCustom TaqMan Advanced Small RNA Assays
Agilent/Stratagene MX3005PN/ASingle tubeCustomUniversal RT1allCustom TaqMan Advanced Small RNA Assays
Agilent/Stratagene MX4000N/ASingle tubeCustomUniversal RT1allCustom TaqMan Advanced Small RNA Assays
TaKaRa T800N/ASingle tubeCustomUniversal RT1allCustom TaqMan Advanced Small RNA Assays
7300 Real-Time PCR System96-wellstdPredesignedUniversal RT1ControlTaqMan™ Advanced miRNA Human Endogenous Controls 96-well Plate, standard
7500 Real-Time PCR System96-wellstdPredesignedUniversal RT1ControlTaqMan™ Advanced miRNA Human Endogenous Controls 96-well Plate, standard
7900HT Real-Time PCR System96-wellstdPredesignedUniversal RT1ControlTaqMan™ Advanced miRNA Human Endogenous Controls 96-well Plate, standard
QuantStudio 3 System96-wellstdPredesignedUniversal RT1ControlTaqMan™ Advanced miRNA Human Endogenous Controls 96-well Plate, standard
QuantStudio 5 System96-wellstdPredesignedUniversal RT1ControlTaqMan™ Advanced miRNA Human Endogenous Controls 96-well Plate, standard
QuantStudio 6 Flex System96-wellstdPredesignedUniversal RT1ControlTaqMan™ Advanced miRNA Human Endogenous Controls 96-well Plate, standard
QuantStudio 7 Flex System96-wellstdPredesignedUniversal RT1ControlTaqMan™ Advanced miRNA Human Endogenous Controls 96-well Plate, standard
QuantStudio 12K Flex System96-wellstdPredesignedUniversal RT1ControlTaqMan™ Advanced miRNA Human Endogenous Controls 96-well Plate, standard
ViiA7 System96-wellstdPredesignedUniversal RT1ControlTaqMan™ Advanced miRNA Human Endogenous Controls 96-well Plate, standard
7500 Fast Real-Time PCR System96-well fast96-well Fast PlatePredesignedUniversal RT1ControlTaqMan™ Advanced miRNA Human Endogenous Controls 96-well Plate, fast
7900HT Real-Time PCR System96-wellfastPredesignedUniversal RT1ControlTaqMan™ Advanced miRNA Human Endogenous Controls 96-well Plate, fast
QuantStudio 3 System96-well fast96-well Fast PlatePredesignedUniversal RT1ControlTaqMan™ Advanced miRNA Human Endogenous Controls 96-well Plate, fast
QuantStudio 5 System96-well fast96-well Fast PlatePredesignedUniversal RT1ControlTaqMan™ Advanced miRNA Human Endogenous Controls 96-well Plate, fast
QuantStudio 6 Flex System96-well fast96-well Fast PlatePredesignedUniversal RT1ControlTaqMan™ Advanced miRNA Human Endogenous Controls 96-well Plate, fast
QuantStudio 7 Flex System96-well fast96-well Fast PlatePredesignedUniversal RT1ControlTaqMan™ Advanced miRNA Human Endogenous Controls 96-well Plate, fast
QuantStudio 12K Flex System96-well fast96-well Fast PlatePredesignedUniversal RT1ControlTaqMan™ Advanced miRNA Human Endogenous Controls 96-well Plate, fast
StepOnePlus System96-well fast96-well Fast PlatePredesignedUniversal RT1ControlTaqMan™ Advanced miRNA Human Endogenous Controls 96-well Plate, fast
ViiA7 System96-well fast96-well Fast PlatePredesignedUniversal RT1ControlTaqMan™ Advanced miRNA Human Endogenous Controls 96-well Plate, fast
Bio-Rad CFX9696-well fast96-well Fast PlatePredesignedUniversal RT1ControlTaqMan™ Advanced miRNA Human Endogenous Controls 96-well Plate, fast
7300 Real-Time PCR System96-well fast96-well Standard PlatePredesignedUniversal RT1AllTaqMan™ Advanced miRNA Human Fixed Content 96-well Plate, standard
7500 Real-Time PCR System96-well fast96-well Standard PlatePredesignedUniversal RT1AllTaqMan™ Advanced miRNA Human Fixed Content 96-well Plate, standard
7900HT Real-Time PCR System96-well fast96-well Standard PlatePredesignedUniversal RT1AllTaqMan™ Advanced miRNA Human Fixed Content 96-well Plate, standard
QuantStudio 3 System96-well fast96-well Standard PlatePredesignedUniversal RT1AllTaqMan™ Advanced miRNA Human Fixed Content 96-well Plate, standard
QuantStudio 5 System96-well fast96-well Standard PlatePredesignedUniversal RT1AllTaqMan™ Advanced miRNA Human Fixed Content 96-well Plate, standard
QuantStudio 6 Flex System96-well fast96-well Standard PlatePredesignedUniversal RT1AllTaqMan™ Advanced miRNA Human Fixed Content 96-well Plate, standard
QuantStudio 7 Flex System96-well fast96-well Standard PlatePredesignedUniversal RT1AllTaqMan™ Advanced miRNA Human Fixed Content 96-well Plate, standard
QuantStudio 12K Flex System96-well fast96-well Standard PlatePredesignedUniversal RT1AllTaqMan™ Advanced miRNA Human Fixed Content 96-well Plate, standard
ViiA7 System96-well fast96-well Standard PlatePredesignedUniversal RT1AllTaqMan™ Advanced miRNA Human Fixed Content 96-well Plate, standard
7500 Fast Real-Time PCR System96-well fast96-well Fast PlatePredesignedUniversal RT1AllTaqMan™ Advanced miRNA Human Fixed Content 96-well Plate, fast
7900HT Real-Time PCR System96-well fast96-well Fast PlatePredesignedUniversal RT1AllTaqMan™ Advanced miRNA Human Fixed Content 96-well Plate, fast
QuantStudio 3 System96-well fast96-well Fast PlatePredesignedUniversal RT1AllTaqMan™ Advanced miRNA Human Fixed Content 96-well Plate, fast
QuantStudio 5 System96-well fast96-well Fast PlatePredesignedUniversal RT1AllTaqMan™ Advanced miRNA Human Fixed Content 96-well Plate, fast
QuantStudio 6 Flex System96-well fast96-well Fast PlatePredesignedUniversal RT1AllTaqMan™ Advanced miRNA Human Fixed Content 96-well Plate, fast
QuantStudio 7 Flex System96-well fast96-well Fast PlatePredesignedUniversal RT1AllTaqMan™ Advanced miRNA Human Fixed Content 96-well Plate, fast
QuantStudio 12K Flex System96-well fast96-well Fast PlatePredesignedUniversal RT1AllTaqMan™ Advanced miRNA Human Fixed Content 96-well Plate, fast
StepOnePlus System96-well fast96-well Fast PlatePredesignedUniversal RT1AllTaqMan™ Advanced miRNA Human Fixed Content 96-well Plate, fast
ViiA7 System96-well fast96-well Fast PlatePredesignedUniversal RT1AllTaqMan™ Advanced miRNA Human Fixed Content 96-well Plate, fast
Bio-Rad CFX9696-well fast96-well Fast PlatePredesignedUniversal RT1AllTaqMan™ Advanced miRNA Human Fixed Content 96-well Plate, fast
7900HT Real-Time PCR SystemTaqMan Array384 well microfluidic cardPredesignedUniversal RT1Pre-Configured Content TaqMan Advanced miRNA Cards
QuantStudio 6 Flex SystemTaqMan Array384 well microfluidic cardPredesignedUniversal RT1Pre-Configured Content TaqMan Advanced miRNA Cards
QuantStudio 7 Flex System TaqMan Array384 well microfluidic cardPredesignedUniversal RT1Pre-Configured Content TaqMan Advanced miRNA Cards
QuantStudio 12K Flex SystemTaqMan Array384 well microfluidic cardPredesignedUniversal RT1Pre-Configured Content TaqMan Advanced miRNA Cards
ViiA7 SystemTaqMan Array384 well microfluidic cardPredesignedUniversal RT1Pre-Configured Content TaqMan Advanced miRNA Cards
7900HT Real-Time PCR SystemTaqMan Array384 well microfluidic cardPredesigned or customUniversal RT10Custom Content TaqMan Advanced miRNA Cards
QuantStudio 6 Flex SystemTaqMan Array384 well microfluidic cardPredesigned or customUniversal RT10Custom Content TaqMan Advanced miRNA Cards
QuantStudio 7 Flex System TaqMan Array384 well microfluidic cardPredesigned or customUniversal RT10Custom Content TaqMan Advanced miRNA Cards
QuantStudio 12K Flex SystemTaqMan Array384 well microfluidic cardPredesigned or customUniversal RT10Custom Content TaqMan Advanced miRNA Cards
ViiA7 SystemTaqMan Array384 well microfluidic cardPredesigned or customUniversal RT10Custom Content TaqMan Advanced miRNA Cards
QuantStudio 12K Flex SystemOpenArray3072 Through holesPredesignedUniversal RT1TaqMan™ OpenArray™ Human Advanced MicroRNA Panel, QuantStudio™ 12K Flex
QuantStudio 12K Flex SystemOpenArray3072 Through holesPredesignedUniversal RT1TaqMan™ OpenArray™ Rodent Advanced MicroRNA Panel, QuantStudio™ 12K Flex
For Research Use Only. Not for use in diagnostic procedures.