Search Thermo Fisher Scientific
Select your dry bath from the filters below to see which blocks are compatible. Additionally, you can take the time to review our available Thermo Scientific Dry Bath portfolio as well.
Product: Flat bottom, 46 x 6mm, 31mm depth SKU: 88870101 Max Block Capacity: 46 | ||
---|---|---|
![]() | Compatible Dry Bath: Digital/Touchscreen | Product Type: Block |
Supported Format: Tubes | Depth mm: 31 | |
Bottom: Flat | Tube Volume: NA | |
Max Block Capacity: 46 | Hole Diameter: 6mm | |
Shaking Speed: NA | Block WxDxH mm/in: 124 x 76 x 39 / 4.9 x 3.0 x 1.5 |
Product: Flat bottom, 28 x 10mm, 31mm depth SKU: 88870102 Max Block Capacity: 28 | ||
---|---|---|
![]() | Compatible Dry Bath: Digital/Touchscreen | Product Type: Block |
Supported Format: Tubes | Depth mm: 31 | |
Bottom: Conical | Tube Volume: NA | |
Max Block Capacity: 28 | Hole Diameter: 10mm | |
Shaking Speed: NA | Block WxDxH mm/in: 124 x 76 x 39 / 4.9 x 3.0 x 1.5 |
Product: Conical bottom, 28 x 1.5mL, 10.8mm, 31mm depth SKU: 88870103 Max Block Capacity: 28 | ||
---|---|---|
![]() | Compatible Dry Bath: Digital/Touchscreen | Product Type: Block |
Supported Format: Tubes | Depth mm: 31 | |
Bottom: Conical | Tube Volume: 1.5mL | |
Max Block Capacity: 28 | Hole Diameter: 10.8mm | |
Shaking Speed: NA | Block WxDxH mm/in: 124 x 76 x 39 / 4.9 x 3.0 x 1.5 |
Product: Conical bottom, 28 x 2.0mL, 10.8mm, 31mm depth SKU: 88870104 Max Block Capacity: 28 | ||
---|---|---|
![]() | Compatible Dry Bath: Digital/Touchscreen | Product Type: Block |
Supported Format: Tubes | Depth mm: 31 | |
Bottom: Conical | Tube Volume: 2.0mL | |
Max Block Capacity: 28 | Hole Diameter: 10.8mm | |
Shaking Speed: NA | Block WxDxH mm/in: 124 x 76 x 39 / 4.9 x 3.0 x 1.5 |
Product: Flat bottom, 28 x 13mm, 31mm depth SKU: 88870105 Max Block Capacity: 24 | ||
---|---|---|
![]() | Compatible Dry Bath: Digital/Touchscreen | Product Type: Block |
Supported Format: Tubes | Depth mm: 31 | |
Bottom: Flat | Tube Volume: NA | |
Max Block Capacity: 24 | Hole Diameter: 13mm | |
Shaking Speed: NA | Block WxDxH mm/in: 124 x 76 x 39 / 4.9 x 3.0 x 1.5 |
Product: Flat bottom, 15 x 16mm, 31mm depth SKU: 88870106 Max Block Capacity: 15 | ||
---|---|---|
![]() | Compatible Dry Bath: Digital/Touchscreen | Product Type: Block |
Supported Format: Tubes | Depth mm: 31 | |
Bottom: Flat | Tube Volume: NA | |
Max Block Capacity: 15 | Hole Diameter: 16mm | |
Shaking Speed: NA | Block WxDxH mm/in: 124 x 76 x 39 / 4.9 x 3.0 x 1.5 |
Product: Flat bottom, 12 x 18mm, 31mm depth SKU: 88870107 Max Block Capacity: 12 | ||
---|---|---|
![]() | Compatible Dry Bath: Digital/Touchscreen | Product Type: Block |
Supported Format: Tubes | Depth mm: 31 | |
Bottom: Flat | Tube Volume: NA | |
Max Block Capacity: 12 | Hole Diameter: 18mm | |
Shaking Speed: NA | Block WxDxH mm/in: 124 x 76 x 39 / 4.9 x 3.0 x 1.5 |
Product: Flat bottom, 8 x 20mm, 31mm depth SKU: 88870108 Max Block Capacity: 8 | ||
---|---|---|
![]() | Compatible Dry Bath: Digital/Touchscreen | Product Type: Block |
Supported Format: Tubes | Depth mm: 31 | |
Bottom: Flat | Tube Volume: NA | |
Max Block Capacity: 8 | Hole Diameter: 20mm | |
Shaking Speed: NA | Block WxDxH mm/in: 124 x 76 x 39 / 4.9 x 3.0 x 1.5 |
Product: Flat bottom, 6 x 25mm, 31mm depth SKU: 88870109 Max Block Capacity: 6 | ||
---|---|---|
![]() | Compatible Dry Bath: Digital/Touchscreen | Product Type: Block |
Supported Format: Tubes | Depth mm: 31 | |
Bottom: Flat | Tube Volume: NA | |
Max Block Capacity: 6 | Hole Diameter: 25mm | |
Shaking Speed: NA | Block WxDxH mm/in: 124 x 76 x 39 / 4.9 x 3.0 x 1.5 |
Product: Conical bottom, 40 x 0.5mL, 8 mm, 21mm depth SKU: 88870110 Max Block Capacity: 40 | ||
---|---|---|
![]() | Compatible Dry Bath: Digital/Touchscreen | Product Type: Block |
Supported Format: Tubes | Depth mm: 21 | |
Bottom: Conical | Tube Volume: 0.5mL | |
Max Block Capacity: 40 | Hole Diameter: 8mm | |
Shaking Speed: NA | Block WxDxH mm/in: 124 x 76 x 39 / 4.9 x 3.0 x 1.5 |
Product: 32 x 6mm & 21 x 10mm, 31 mm depth SKU: 88870112 Max Block Capacity: 32 + 21 | ||
---|---|---|
![]() | Compatible Dry Bath: Digital/Touchscreen | Product Type: Block |
Supported Format: Mixed tubes | Depth mm: 31 | |
Bottom: Flat | Tube Volume: NA | |
Max Block Capacity: 32 + 21 | Hole Diameter: 6mm / 10mm | |
Shaking Speed: NA | Block WxDxH mm/in: 124 x 76 x 39 / 4.9 x 3.0 x 1.5 |
Product: 18 x 1.5mL & 10 x 2.0mL SKU: 88870113 Max Block Capacity: 18 + 10 | ||
---|---|---|
![]() | Compatible Dry Bath: Digital/Touchscreen | Product Type: Block |
Supported Format: Mixed tubes | Depth mm: 31 | |
Bottom: Conical | Tube Volume: 2.0mL / 1.5mL | |
Max Block Capacity: 18 + 10 | Hole Diameter: 10.8mm | |
Shaking Speed: NA | Block WxDxH mm/in: 124 x 76 x 39 / 4.9 x 3.0 x 1.5 |
Product: Mixed block, 3 x 25mm & 12 x 12~13mm & 6 x 6mm, 31 mm depth SKU: 88870114 Max Block Capacity: 3 + 12 + 6 | ||
---|---|---|
![]() | Compatible Dry Bath: Digital/Touchscreen | Product Type: Block |
Supported Format: Mixed tubes | Depth mm: 31 | |
Bottom: Flat | Tube Volume: NA | |
Max Block Capacity: 3 + 12 + 6 | Hole Diameter: 25mm / 13mm / 6mm | |
Shaking Speed: NA | Block WxDxH mm/in: 124 x 76 x 39 / 4.9 x 3.0 x 1.5 |
Product: Mixed block, 30 x 0.5mL & 20 x 0.2mL, 31mm depth SKU: 88870115 Max Block Capacity: 30 + 20 | ||
---|---|---|
![]() | Compatible Dry Bath: Digital/Touchscreen | Product Type: Block |
Supported Format: Mixed tubes | Depth mm: 31 | |
Bottom: Conical | Tube Volume: 0.5mL / 0.2mL | |
Max Block Capacity: 30 + 20 | Hole Diameter: 8mm / 5.5mm | |
Shaking Speed: NA | Block WxDxH mm/in: 124 x 76 x 39 / 4.9 x 3.0 x 1.5 |
Product: Flat bottom, 15 x 15mL, 17mm, 115mm depth SKU: 88870116 Max Block Capacity: 15 | ||
---|---|---|
![]() | Compatible Dry Bath: Digital/Touchscreen | Product Type: Block |
Supported Format: Tubes | Depth mm: 115 | |
Bottom: Flat | Tube Volume: 15mL | |
Max Block Capacity: 15 | Hole Diameter: 17mm | |
Shaking Speed: NA | Block WxDxH mm/in: 124 x 76 x 39 / 4.9 x 3.0 x 1.5 |
Product: Flat bottom, 5 x 50mL, 29.8mm, 115mm depth SKU: 88870117 Max Block Capacity: 4 | ||
---|---|---|
![]() | Compatible Dry Bath: Digital/Touchscreen | Product Type: Block |
Supported Format: Tubes | Depth mm: 115 | |
Bottom: Flat | Tube Volume: 50mL | |
Max Block Capacity: 4 | Hole Diameter: 29.8mm | |
Shaking Speed: NA | Block WxDxH mm/in: 124 x 76 x 39 / 4.9 x 3.0 x 1.5 |
Product: Conical bottom, 15 x 15mL, 17mm, 115mm depth SKU: 88870118 Max Block Capacity: 15 | ||
---|---|---|
![]() | Compatible Dry Bath: Digital/Touchscreen | Product Type: Block |
Supported Format: Tubes | Depth mm: 115 | |
Bottom: Conical | Tube Volume: 15mL | |
Max Block Capacity: 15 | Hole Diameter: 17mm | |
Shaking Speed: NA | Block WxDxH mm/in: 124 x 76 x 39 / 4.9 x 3.0 x 1.5 |
Product: Conical bottom, 4 x 50mL, 29.8mm, 115mm depth SKU: 88870119 Max Block Capacity: 4 | ||
---|---|---|
![]() | Compatible Dry Bath: Digital/Touchscreen | Product Type: Block |
Supported Format: Tubes | Depth mm: 115 | |
Bottom: Conical | Tube Volume: 50mL | |
Max Block Capacity: 4 | Hole Diameter: 29.8mm | |
Shaking Speed: NA | Block WxDxH mm/in: 124 x 76 x 39 / 4.9 x 3.0 x 1.5 |
Product: Conical bottom, 1 x 96-well 0.2mL, non-skirted PCR Plate, 11mm depth SKU: 88870120 Max Block Capacity: 1 | ||
---|---|---|
![]() | Compatible Dry Bath: Digital/Touchscreen | Product Type: Block |
Supported Format: Plate | Depth mm: 11 | |
Bottom: Conical | Tube Volume: 0.2mL | |
Max Block Capacity: 1 | Hole Diameter: 5.5mm | |
Shaking Speed: NA | Block WxDxH mm/in: 124 x 76 x 39 / 4.9 x 3.0 x 1.5 |
Product: Temperature probe PT1000, stainless steel (Only compatible with Touchscreen dry baths) SKU: 88870122 Max Block Capacity: NA | ||
---|---|---|
![]() | Compatible Dry Bath: Digital/Touchscreen | Product Type: Temp. Probe |
Supported Format: NA | Depth mm: NA | |
Bottom: NA | Tube Volume: NA | |
Max Block Capacity: NA | Hole Diameter: NA | |
Shaking Speed: NA | Block WxDxH mm/in: NA |
Product: Block Lifter SKU: 88880123 Max Block Capacity: NA | ||
---|---|---|
![]() | Compatible Dry Bath: Digital/Touchscreen | Product Type: Block Lifter |
Supported Format: NA | Depth mm: NA | |
Bottom: NA | Tube Volume: NA | |
Max Block Capacity: NA | Hole Diameter: NA | |
Shaking Speed: NA | Block WxDxH mm/in: NA |
Product: Block Lifter SKU: 88870123 Max Block Capacity: NA | ||
---|---|---|
![]() | Compatible Dry Bath: Digital/Touchscreen | Product Type: Block Lifter |
Supported Format: NA | Depth mm: NA | |
Bottom: NA | Tube Volume: NA | |
Max Block Capacity: NA | Hole Diameter: NA | |
Shaking Speed: NA | Block WxDxH mm/in: NA |
Product: 0.2ml quarter block (Max: Dry Bath S 4 blocks, Dry Bath D 8 blocks) SKU: 88871101 Max Block Capacity: 24 | ||
---|---|---|
![]() | Compatible Dry Bath: Compact | Product Type: Block |
Supported Format: Tubes | Depth mm: 13 | |
Bottom: Conical | Tube Volume: 0.2mL | |
Max Block Capacity: 24 | Hole Diameter: 6mm | |
Shaking Speed: NA | Block WxDxH mm/in: 54 x 36 x 15 / 2.1 x 1.4 x 0.6 |
Product: 0.5ml quarter block (Max: Dry Bath S 4 blocks, Dry Bath D 8 blocks) SKU: 88871102 Max Block Capacity: 12 | ||
---|---|---|
![]() | Compatible Dry Bath: Compact | Product Type: Block |
Supported Format: Tubes | Depth mm: 25 | |
Bottom: Conical | Tube Volume: 0.5mL | |
Max Block Capacity: 12 | Hole Diameter: 7.8mm | |
Shaking Speed: NA | Block WxDxH mm/in: 54 x 36 x 30 / 2.1 x 1.4 x 1.2 |
Product: 1.5ml quarter block (Max: Dry Bath S 4 blocks, Dry Bath D 8 blocks) SKU: 88871103 Max Block Capacity: 6 | ||
---|---|---|
![]() | Compatible Dry Bath: Compact | Product Type: Block |
Supported Format: Tubes | Depth mm: 27 | |
Bottom: Conical | Tube Volume: 1.5mL | |
Max Block Capacity: 6 | Hole Diameter: 10.8mm | |
Shaking Speed: NA | Block WxDxH mm/in: 54 x 36 x 30 / 2.1 x 1.4 x 1.2 |
Product: 2.0ml quarter block (Max: Dry Bath S 4 blocks, Dry Bath D 8 blocks) SKU: 88871104 Max Block Capacity: 6 | ||
---|---|---|
![]() | Compatible Dry Bath: Compact | Product Type: Block |
Supported Format: Tubes | Depth mm: 27 | |
Bottom: Flat | Tube Volume: 2.0mL | |
Max Block Capacity: 6 | Hole Diameter: 11mm | |
Shaking Speed: NA | Block WxDxH mm/in: 54 x 36 x 30 / 2.1 x 1.4 x 1.2 |
Product: 15ml half block (Max: Dry Bath S 2 blocks, Dry Bath D 4 blocks) SKU: 88871105 Max Block Capacity: 6 | ||
---|---|---|
![]() | Compatible Dry Bath: Compact | Product Type: Block |
Supported Format: Tubes | Depth mm: 69 | |
Bottom: Flat | Tube Volume: 15mL | |
Max Block Capacity: 6 | Hole Diameter: 16.5mm | |
Shaking Speed: NA | Block WxDxH mm/in: 108 x 36 x 72 / 4.3 x 1.4 x 2.8 |
Product: 50 ml half block (Max: Dry Bath S 2 blocks, Dry Bath D 4 blocks) SKU: 88871106 Max Block Capacity: 3 | ||
---|---|---|
![]() | Compatible Dry Bath: Compact | Product Type: Block |
Supported Format: Tubes | Depth mm: 69 | |
Bottom: Flat | Tube Volume: 50mL | |
Max Block Capacity: 3 | Hole Diameter: 29.5mm | |
Shaking Speed: NA | Block WxDxH mm/in: 108 x 36 x 72 / 4.3 x 1.4 x 2.8 |
Product: Heat insulator for Compact S (2) SKU: 88871108 Max Block Capacity: NA | ||
---|---|---|
![]() | Compatible Dry Bath: Compact | Product Type: Heat Insulator |
Supported Format: NA | Depth mm: NA | |
Bottom: NA | Tube Volume: NA | |
Max Block Capacity: NA | Hole Diameter: NA | |
Shaking Speed: NA | Block WxDxH mm/in: 143 x 107 x 80 (40 x 2) / 5.6 x 4.2 x 3.2 (1.6 x 2) |
Product: Heat insulator for Compact D (2) SKU: 88871109 Max Block Capacity: NA | ||
---|---|---|
![]() | Compatible Dry Bath: Compact | Product Type: Heat Insulator |
Supported Format: NA | Depth mm: NA | |
Bottom: NA | Tube Volume: NA | |
Max Block Capacity: NA | Hole Diameter: NA | |
Shaking Speed: NA | Block WxDxH mm/in: 179 x 143 x 80 (40 x 2) / 7.0 x 5.6 x 3.2 (1.6 x 2) |
Product: Plastic lid for Compact S SKU: 88871110 Max Block Capacity: NA | ||
---|---|---|
![]() | Compatible Dry Bath: Compact | Product Type: Lid |
Supported Format: NA | Depth mm: NA | |
Bottom: NA | Tube Volume: NA | |
Max Block Capacity: NA | Hole Diameter: NA | |
Shaking Speed: NA | Block WxDxH mm/in: 122 x 86 x 40 / 4.8 x 3.4 x 1.6 |
Product: Plastic lid for Compact D SKU: 88871111 Max Block Capacity: NA | ||
---|---|---|
![]() | Compatible Dry Bath: Compact | Product Type: Lid |
Supported Format: NA | Depth mm: NA | |
Bottom: NA | Tube Volume: NA | |
Max Block Capacity: NA | Hole Diameter: NA | |
Shaking Speed: NA | Block WxDxH mm/in: 158 x 122 x 40 / 6.2 x 4.8 x 1.6 |
Product: 0.2ml Microplate Block Set with Cover* SKU: 88880125 Max Block Capacity: 96 | ||
---|---|---|
![]() | Compatible Dry Bath: Heating/Shaking | Product Type: Block |
Supported Format: Tubes | Depth mm: 11 | |
Bottom: Conical | Tube Volume: 0.2mL | |
Max Block Capacity: 96 | Hole Diameter: 6mm | |
Shaking Speed: 150-1500 | Block WxDxH mm/in: 111 x 76.5 x 15 / 4.4 x 3 x 0.6 |
Product: 0.5ml Tube Block Set with Cover* SKU: 88880126 Max Block Capacity: 48 | ||
---|---|---|
![]() | Compatible Dry Bath: Heating/Shaking | Product Type: Block |
Supported Format: Tubes | Depth mm: 21 | |
Bottom: Flat | Tube Volume: 0.5mL | |
Max Block Capacity: 48 | Hole Diameter: 7.8mm | |
Shaking Speed: 150-1000 | Block WxDxH mm/in: 98 x 76.5 x 30 / 3.9 x 3 x 1.2 |
Product: 1.5/2.0ml Tube Block Set with Cover* SKU: 88880127 Max Block Capacity: 24 | ||
---|---|---|
![]() | Compatible Dry Bath: Heating/Shaking | Product Type: Block |
Supported Format: Tubes | Depth mm: 30 | |
Bottom: Flat | Tube Volume: 2.0mL / 1.5mL | |
Max Block Capacity: 24 | Hole Diameter: 10.8mm | |
Shaking Speed: 150-1000 | Block WxDxH mm/in: 98 x 76.5 x 41 / 3.9 x 3 x 1.6 |
Product: 50ml Tube Block Set with Cover* SKU: 88880128 Max Block Capacity: 6 | ||
---|---|---|
![]() | Compatible Dry Bath: Heating/Shaking | Product Type: Block |
Supported Format: Tubes | Depth mm: 48 | |
Bottom: Flat | Tube Volume: 50mL | |
Max Block Capacity: 6 | Hole Diameter: 29mm | |
Shaking Speed: 150-800 | Block WxDxH mm/in: 98 x 76.5 x 51 / 3.9 x 3 x 2 |
Product: 15ml Tube Block Set with Cover* SKU: 88880151 Max Block Capacity: 15 | ||
---|---|---|
![]() | Compatible Dry Bath: Heating/Shaking | Product Type: Block |
Supported Format: Tubes | Depth mm: 48 | |
Bottom: Flat | Tube Volume: 15mL | |
Max Block Capacity: 15 | Hole Diameter: 17mm | |
Shaking Speed: 150-900 | Block WxDxH mm/in: 98 x 76.5 x 51 / 3.9 x 3 x 2 |
Product: 13mm dia Tube Block Set with Cover* SKU: 88880153 Max Block Capacity: 24 | ||
---|---|---|
![]() | Compatible Dry Bath: Heating/Shaking | Product Type: Block |
Supported Format: Tubes | Depth mm: 48 | |
Bottom: Flat | Tube Volume: NA | |
Max Block Capacity: 24 | Hole Diameter: 13mm | |
Shaking Speed: 150-850 | Block WxDxH mm/in: 98 x 76.5 x 51 / 3.9 x 3 x 2 |
Product: Block for microtube 0.5ml SKU: 88880129 Max Block Capacity: 48 | ||
---|---|---|
![]() | Compatible Dry Bath: Heating/Cooling | Product Type: Block |
Supported Format: Tubes | Depth mm: 26 | |
Bottom: Flat | Tube Volume: 0.5mL | |
Max Block Capacity: 48 | Hole Diameter: 7.8mm | |
Shaking Speed: NA | Block WxDxH mm/in: 98 x 76.5 x 41 / 3.9 x 3 x 1.6 |
Product: Block for microtube 1.5ml SKU: 88880130 Max Block Capacity: 30 | ||
---|---|---|
![]() | Compatible Dry Bath: Heating/Cooling | Product Type: Block |
Supported Format: Tubes | Depth mm: 35 | |
Bottom: Flat | Tube Volume: 1.5mL | |
Max Block Capacity: 30 | Hole Diameter: 10.8mm | |
Shaking Speed: NA | Block WxDxH mm/in: 98 x 76.5 x 41 / 3.9 x 3 x 1.6 |
Product: Block for centrifuge tube 15ml (Lid will not close) SKU: 88880131 Max Block Capacity: 15 | ||
---|---|---|
![]() | Compatible Dry Bath: Heating/Cooling | Product Type: Block |
Supported Format: Tubes | Depth mm: 48 | |
Bottom: Flat | Tube Volume: 15mL | |
Max Block Capacity: 15 | Hole Diameter: 16.5mm | |
Shaking Speed: NA | Block WxDxH mm/in: 98 x 76.5 x 51 / 3.9 x 3 x 2 |
Product: Short block for centrifuge tube 50ml (Lid will not close) SKU: 88880132 Max Block Capacity: 6 | ||
---|---|---|
![]() | Compatible Dry Bath: Heating/Cooling | Product Type: Block |
Supported Format: Tubes | Depth mm: 48 | |
Bottom: Flat | Tube Volume: 50mL | |
Max Block Capacity: 6 | Hole Diameter: 29mm | |
Shaking Speed: NA | Block WxDxH mm/in: 98 x 76.5 x 51 / 3.9 x 3 x 2 |
Product: Long block for centrifuge tube 50ml (Lid will not close) SKU: 88880133 Max Block Capacity: 6 | ||
---|---|---|
![]() | Compatible Dry Bath: Heating/Cooling | Product Type: Block |
Supported Format: Tubes | Depth mm: 85 | |
Bottom: Flat | Tube Volume: 50mL | |
Max Block Capacity: 6 | Hole Diameter: 29.4mm | |
Shaking Speed: NA | Block WxDxH mm/in: 98 x 76.5 x 87 / 3.9 x 3 x 3.4 |
Product: Block for 10mm tube (Lid will not close) SKU: 88880134 Max Block Capacity: 35 | ||
---|---|---|
![]() | Compatible Dry Bath: Heating/Cooling | Product Type: Block |
Supported Format: Tubes | Depth mm: 48 | |
Bottom: Flat | Tube Volume: NA | |
Max Block Capacity: 35 | Hole Diameter: 10mm | |
Shaking Speed: NA | Block WxDxH mm/in: 98 x 76.5 x 51 / 3.9 x 3 x 2 |
Product: Block for 12mm tube (Lid will not close) SKU: 88880135 Max Block Capacity: 24 | ||
---|---|---|
![]() | Compatible Dry Bath: Heating/Cooling | Product Type: Block |
Supported Format: Tubes | Depth mm: 48 | |
Bottom: Flat | Tube Volume: NA | |
Max Block Capacity: 24 | Hole Diameter: 12mm | |
Shaking Speed: NA | Block WxDxH mm/in: 98 x 76.5 x 51 / 3.9 x 3 x 2 |
Product: Block for 13mm tube (Lid will not close) SKU: 88880136 Max Block Capacity: 24 | ||
---|---|---|
![]() | Compatible Dry Bath: Heating/Cooling | Product Type: Block |
Supported Format: Tubes | Depth mm: 48 | |
Bottom: Flat | Tube Volume: NA | |
Max Block Capacity: 24 | Hole Diameter: 13mm | |
Shaking Speed: NA | Block WxDxH mm/in: 98 x 76.5 x 51 / 3.9 x 3 x 2 |
Product: Block for 16mm tube (Lid will not close) SKU: 88880138 Max Block Capacity: 16 | ||
---|---|---|
![]() | Compatible Dry Bath: Heating/Cooling | Product Type: Block |
Supported Format: Tubes | Depth mm: 48 | |
Bottom: Flat | Tube Volume: NA | |
Max Block Capacity: 16 | Hole Diameter: 16mm | |
Shaking Speed: NA | Block WxDxH mm/in: 98 x 76.5 x 51 / 3.9 x 3 x 2 |
Product: Block for 18mm tube (Lid will not close) SKU: 88880139 Max Block Capacity: 12 | ||
---|---|---|
![]() | Compatible Dry Bath: Heating/Cooling | Product Type: Block |
Supported Format: Tubes | Depth mm: 48 | |
Bottom: Flat | Tube Volume: NA | |
Max Block Capacity: 12 | Hole Diameter: 18mm | |
Shaking Speed: NA | Block WxDxH mm/in: 98 x 76.5 x 51 / 3.9 x 3 x 2 |
Product: Block for 20mm tube (Lid will not close) SKU: 88880140 Max Block Capacity: 12 | ||
---|---|---|
![]() | Compatible Dry Bath: Heating/Cooling | Product Type: Block |
Supported Format: Tubes | Depth mm: 48 | |
Bottom: Flat | Tube Volume: NA | |
Max Block Capacity: 12 | Hole Diameter: 20mm | |
Shaking Speed: NA | Block WxDxH mm/in: 98 x 76.5 x 51 / 3.9 x 3 x 2 |