Live and dead bacteria visualized on freshly isolated human cheek epithelial cells using our LIVE/DEAD® BacLight™ Bacterial Viability Kit.
Live and dead bacteria visualized on freshly isolated human cheek epithelial cells using our LIVE/DEAD® BacLight™ Bacterial Viability Kit (Cat. no. L7007, L7012, L13152). When incubated with the SYTO® 9 and propidium iodide nucleic acid stains provided in this kit, live bacteria with intact cell membranes fluoresce green and dead bacteria with compromised membranes fluoresce red. This image was photographed in a single exposure through an Omega Optical triple bandpass filter set.
Live and dead bacteria visualized on freshly isolated human cheek epithelial cells using our LIVE/DEAD® <i>Bac</i>Light™ Bacterial Viability Kit (Cat. no. L7007, L7012, L13152).  When incubated with the SYTO® 9 and propidium iodide nucleic acid stains provided in this kit, live bacteria with intact cell membranes fluoresce green and dead bacteria with compromised membranes fluoresce red.  This image was photographed in a single exposure through an Omega Optical triple bandpass filter set.
Related Products
Catalog # Name Size Price (USD) Qty
L13152 LIVE/DEAD™ BacLight™ Bacterial Viability Kit, 10 Applicator Sets kit Each
486.65
Online Exclusive
500.00
Save 13.35 (3%)
L7007 LIVE/DEAD™ BacLight™ Bacterial Viability Kit, for microscopy, 1 kit Each
836.65
Online Exclusive
862.00
Save 25.35 (3%)
L7012 LIVE/DEAD™ BacLight™ Bacterial Viability Kit, for microscopy & quantitative assays, 1 kit Each
607.65
Online Exclusive
626.00
Save 18.35 (3%)
Related Images