Fixed and permeabilized bovine pulmonary artery endothelial cells labeled with the F-actin stain, fluorescein phalloidin, followed by an Alexa Fluor 594 anti–fluorescein/Oregon Green rabbit IgG antibody.
Fixed and permeabilized bovine pulmonary artery endothelial cells were labeled with the filamentous actin (F-actin) stain, fluorescein phalloidin (F432, right). An Alexa Fluor 594 anti–fluorescein/Oregon Green rabbit IgG antibody (A11091) converted the green fluorescence to red (left).
Fixed and permeabilized bovine pulmonary artery endothelial cells were labeled with the filamentous actin (F-actin) stain, fluorescein phalloidin (F432, right). An Alexa Fluor 594 anti–fluorescein/Oregon Green rabbit IgG antibody (A11091) converted the green fluorescence to red (left).
Related Products
Catalog # Name Size Price (USD) Qty
A11091 Fluorescein/Oregon Green Polyclonal Antibody, Alexa Fluor™ 594 500 µL -
F432 Fluorescein Phalloidin, Green, 496/516 nm Each
586.65
Online Exclusive
606.00
Save 19.35 (3%)
Related Images