Tradeshow
Oct 17, 2023 - Oct 19, 2023
Huntington Place, Detroit, MI, USA
The International Materials, Applications & Technologies (IMAT) meeting, organized by ASM, is focused on advanced materials, applications and technologies addressing a spectrum of emerging technologies in key growth market areas to solve global materials challenges. Visit us in Booth #1520 to talk to our experts about our latest innovations that can advance your materials research.
Featured Systems
Axia ChemiSEM
Unlike traditional scanning electron microscopes, the Axia ChemiSEM constantly processes EDS data in the background, giving you live updates on elemental data, as it is acquired. The Axia ChemiSEM’s workflows put elemental mapping first, not last, and reveal otherwise unseen compositional variation about your samples. Applications encompass metals, additive powders, batteries, and technical ceramics.
Perception Particle Analysis software
Thermo Scientific Perception Automated Particle Analysis Software provides a fully integrated, automated, and user-friendly solution for particle analysis on an SEM. Book a demo today to learn more!
Book a demo to see its key features, such as how elements found in your sample can be toggled on and off, allowing you to isolate areas of interest.
Book a demo ›
Solutions Center presentations:
October 17, 2023
10:50 a.m. EDT
Presenter: John Yorston
In this short presentation, we will look at several examples of how Thermo Scientific ChemiSEM Technology and ChemiPhase are used to accelerate the time to accurate SEM-EDX results.
Scanning electron microscopy and energy dispersive X-ray microanalysis (SEM-EDX) are widely used in the characterization of materials. Advances in computing power, together with the application of adaptive super-pixel clustering algorithms, have vastly decreased the time necessary to map, determine elemental compositions, and delineate statistically similar component phases.
ChemiSEM Technology uses a big data approach to detect all statistically significant spectra within the acquired data cube. ChemiPhase software then provides a simple probability for each pixel, indicating if it belongs to each of these spectra. This makes interpretation of complex samples much more straightforward and intuitive, as each pixel can belong to only a single phase.
John Yorston
October 17, 2023
11:20 a.m. EDT
Presenter: Rogier Miltenburg
Scanning electron microscopy is used not only for research but also for daily quality control. Steel, precision parts, and battery components are routinely inspected for contaminations or defects. Depending on the size, shape, and composition, particles can have a negative impact on the quality of the finished product; therefore, it is critical to understand the properties of the particles. Learn how new Thermo Scientific Perception Software enables advanced SEM-EDS automated particle analysis. Perception Software is an all-in-one software package that can turn your scanning electron microscope of choice into a dedicated solution for your particle analysis needs.
Rogier Miltenburg
Book a demo for IMAT 2023
*
Required field
*
*
*
*
 Speak to a representative
 Schedule a demo
 Request a quote
 Speak to a service representative about our service offerings
 Learn more information as a student
 I would like to receive marketing and promotional emails regarding products and services from Thermo Fisher Scientific.
This site is protected by reCAPTCHA and the Google Privacy Policy and Terms of Service apply.
Style Sheet for Global Design System
Style Sheet for Komodo Tabs