The Fischer Indole Syntheis is the most important method of preparing substituted indoles.
Fischer Indole Synthesis Overview
One of the oldest reactions in organic chemistry, the Fischer Indole synthesis is the condensation of a substituted phenylhydrazine and a carbonyl compound under acidic conditions to produce an aromatic heterocycle indole.
Named Reactions in Organic Synthesis
Fischer Indole Synthesis Mechanism
Watch our video to learn more about the Fischer Indole synthesis and its mechanisms.
Video Player is loading.
Current Time 0:00
Duration 2:53
Loaded: 0%
Stream Type LIVE
Remaining Time 2:53
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Need reagents for the Fischer Indole Synthesis?
    Chem dex
    If you need further help to plan your synthesis, don’t miss our new app.  Chem dex is a new tool to explore hundreds of chemical reactions, and to have access to product recommendations. Visit: thermofisher.com/chemdex.
    Named Reactions eBook
    Named Reactions in Organic Synthesis eBook
    Download our Named Reactions eBook to learn more about the Fischer Indole Synthesis. 
    Access 10 chapters of content about organic chemistry reactions that are named after the chemists that discovered them. This informative eBook includes quizzes so you can test your knowledge!
    History of the Fischer Indole Synthesis
    In 1883, E. Fischer and F. Jourdan treated pyruvic acid 1-methylphenylhydrazone with alcoholic hydrogen chloride and generated 1-methylindole-2-carboxylic acid. Preparing indoles by heating the arylhydrazones of either aldehydes or ketones in the presence of a protic or Lewis acid is now known as the Fischer indole synthesis. Since its discovery, it has remained the most important method of preparing substituted indoles.
    Main features of the Fischer indole synthesis:
    • The indole formation can be carried out as a one-pot synthesis, as it is not necessary to isolate the intermediate arylhydrazones
    • Unsymmetrical ketones give two region-isomeric 2,3-disubstituted indoles, with the region-selectivity dependent on acidity of the medium, substitution of the hydrazine, and steric effects
    • 1,2-diketones can give both mono and bis-indoles, the mono-indoles usually forming with strong acid catalysts in refluxing alcohols
    Other Heterocycle Formation Reactions
    Acroseal Packaging
    Chemical reactions often involve the use of air- and moisture-sensitive solvents, and pyrophoric or hazardous reagents. Our AcroSeal packaging is a packaging solution designed to enable safe handling of these types of materials which are used in a variety of research and development applications, including NMR analysis and studies in drug discovery, agrochemicals, flavors and fragrances, and more. 
    Watch our video for more information. 
    Video Player is loading.
    Current Time 0:00
    Duration 4:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 4:00