Watch this video to learn more about the CryoMed CRF
Video Player is loading.
Current Time 0:00
Duration 0:00
Loaded: 0%
Stream Type LIVE
Remaining Time 0:00
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Riptide-Lab
      Integrated OPC UA protocol within the new CryoMed CRF firmware to enable remote unit communication via DeltaV and other systems*.
      • OPC UA = Open Platform Communications Unified Architecture
      • OPC UA: Machine-to-machine communication protocol for industrial automation
      • DeltaV™: Distributed control system for automation systems
      *OPC UA will open-up additional opportunities utilizing in-house automation and IT teams for integration with a data system.
      product features
      CryoMed consistent freezing results
      Enhanced data traceability and customized freezing profiles for consistent temp. performance and real-time monitoring.
      CTS documentation
      Documentation package
      CryoMed CRF with OPC UA comes equipped with a user manual, a certificate of conformance, and a certificate of calibration. 
      Services flyer
      Compliance services
      Service options include installation qualification (IQ), operation qualification (OQ), and temperature mapping.
      Medical Device (available in North America Only)
      • FDA listed Class II medical device, 510K exempt
      • Available in 17 L, 34 L, 48 L
      • With and without built-in thermal printer
      • 100V
      • 120V
      • 220-230V
      General Purpose ( available everywhere)
      • Available in 17 L, 34 L, 48 L
      • With and without built-in thermal printer
      • 100V
      • 120V
      • 220-230V
      Cryomed 3D Tour
      Note: This tool is best viewed in Chrome, Firefox or Safari Browsers. Learn more ›
      CryoMed-blog
      Blog post
      CryoMed Controlled-Rate Freezers that meet 21 CFR Part 11 requirements: Safeguarding sample integrity, traceability and 21 CFR Part 11 compliance.
      Read more >
      Webinar
      On-demand webinar
      A Partnership in Data Traceability: 21 CFR part 11, the CryoMed controlled-rate freezer, and Your Lab/Production Processes
      Watch on-demand >
      Smart Note
      Smart Note
      How can the CryoMed CRF be used to prepare samples and therapies for long-term storage in a cryogenic storage device?
      Read now >
      Style Sheet for Global Design System