Molecular diagnostic solutions for respiratory infection testing
Prompt and early diagnosis of respiratory pathogens that cause severe illness is critical to help inform patient management, treatment decisions, and preventing the spread of disease.  Confidence in testing results allows clinicians and patients to make informed decisions—including seeking medical care and/or helping to prevent contact with other people. While many testing options are available for respiratory testing, molecular techniques including PCR-based tests are designed to enable early virus detection, even in asymptomatic cases, and help deliver reliable answers.
Webinar: Viruses Circulating in the 2023–2024 Flu Season: What Have We Seen?
TaqPath COVID-19, Flu A, Flu B, RSV Select Panel
The Applied Biosystems TaqPath COVID‑19, Flu A, Flu B, RSV Select Panel is a multiplex, real-time reverse transcription polymerase chain reaction (RT-PCR) in vitro diagnostic test for the qualitative detection and differentiation of severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2), influenza A virus, influenza B virus, and respiratory syncytial virus (RSV).
Components included in the Applied Biosystems TaqPath COVID‑19, Flu A, Flu B, RSV Select Panel
TaqPath COVID-19 Diagnostic PCR Kit
The TaqPath COVID-19 Diagnostic PCR Kit is a real-time reverse transcription polymerase chain reaction (RT-PCR) test intended for the qualitative detection of nucleic acids from SARS-CoV-2 in nasopharyngeal and anterior nasal specimens from individuals with signs and symptoms of respiratory tract infection.
Product image showing all the components included in the TaqPath COVID-19 Diagnostic PCR Kit
TaqPath COVID-19, Flu A, Flu B Combo Kit
Boost your lab's efficiency with the Applied Biosystems TaqPath COVID-19, Flu A, Flu B Combo Kit by Thermo Fisher Scientific. This high-precision multiplex real-time PCR test enables simultaneous detection and differentiation of SARS-CoV-2, influenza A, and influenza B. Designed to accommodate emerging SARS-CoV-2 variants, this kit expands your respiratory sample testing capabilities while maintaining low operational costs. Authorized for use with multiple sample types, it is designed to deliver a turnaround time of approximately 3 hours. The kit includes the Pathogen Interpretive Software, aimed at transforming complex genetic data into easy-to-read reports and minimizing interpretation errors. Enhance your diagnostic accuracy and efficiency with this comprehensive solution.
TaqPath COvid-FluA-FluB kit
For In Vitro Diagnostic Use.