TCAutomation Laboratory Automation System
Efficiency and consistency in sample handling
Expandable automation to streamline lab processes
Improved efficiency and safety of sample management together with short and consistent turnaround times are the key benefits of laboratory automation. By automating the key pre- and post-analytical steps, such as decapping, centrifugation, aliquoting, sorting, recapping, storage and retrieval, Thermo Scientific™ TCAutomation™ helps to manage the increasing workloads and improve quality in clinical laboratories.
Contact us to speak with a distributor
Flexible, open and scalable system
TCAutomation system provides a solution for increased throughput needs and for integration of new analyzing technologies.
Each installation is planned for its individual optimal efficiency delivering improved sample management.
  • TCAutomation systems fit into various floor plans.
  • The modular concepts are scalable to meet the changing needs.
  • TCAutomation system can grow from a small automation to high volume total laboratory automation solution.
TCAutomation system supports analyzer interfaces from various different manufactures. Workcell solutions may include two or more system combination increasing the analyzer capacity and decreasing the manual work.
  • Single entry point without pre-sorting.
  • One centralized user interface offers integrated functionality for managing the whole workcell.
  • Easy addition of pre- and post-analytical functionality.
Watch TCAutomation system in action
TCAutomation BES Flex - Fully automated sample input and sorter system
TCAutomation BES Flex is a fully automated sample input and sorter system for batches of securely closed sample tubes.
Video Player is loading.
Current Time 0:00
Duration 1:17
Loaded: 0%
Stream Type LIVE
Remaining Time 1:17
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
      1. Now Playing
        Up NextTCAutomation BES Flex - Fully automated sample input and sorter system
        TCAutomation BES Flex is a fully automated sample input and sorter system for batches of securely closed sample tubes.
      2. Now Playing
        Up NextTailor-made TCAutomation Systems manufactured to fit into even the most challenging laboratories
        Every ordered TCAutomation System undergoes several test procedures to ensure reliability. At customer laboratory this ensures short installation time.
      3. Now Playing
        Up NextThermo Scientific TCAutomation - Open and Expandable Laboratory Automation Solution
        TCAutomation solution manages clinical laboratories workloads by automating pre- and post-analytical steps and integrating various manufactures analyzers.
      4. Now Playing
        Up NextTCAutomation Conveyors
        Dual lane conveyors in different lengths for sample transport. Each system includes at least two U-Turns. L-, T-, Z- and internal U-Turns also available.
      5. Now Playing
        Up NextTCAutomation Cold Storage System
        Cold Storage is a refrigerated storage system from which samples can be automatically retrieved back to the track for re-testing or discharged according to the programmable timing. Storage capacity for up to 14 800 tubes.
      6. Now Playing
        Up NextTCAutomation Centrifuge Module
        Module for automatic and continuous centrifugation of samples. Automates loading, balancing and unloading of centrifuge racks.
      7. Now Playing
        Up NextTCAutomation Decapper Flex Module
        Module for automatic cap removal from different types and sizes of sample tubes.
      8. Now Playing
        Up NextTCAutomation Recapper Module
        Module for automatic capping of different types and sizes of primary and TCAutomation Aliquoter secondary tubes with conical caps.
      9. Now Playing
        Up NextTCAutomation Aliquoter & Labeler Module
        Module for automatic sample aliquoting to secondary tubes using disposable tips. Automatic barcode labeling of secondary tubes.
      10. Now Playing
        Up NextTCAutomation ES Flex Module
        ES Flex is a module for automatic sample loading and unloading. Module has automatic retrieval for rerun and excellent sample sorting capabilities.
    Labor-intensive tasks can be automated in different combinations
    TCAutomation ES Flex, Entry and Exit Modules
    ES Flex, Entry and Exit Modules - Modules for automatic or manual sample loading and unloading in user-specified racks. In ES Flex automatic retrieval for rerun and excellent sample sorting capabilities.
    TCAutomation Aliquoter/Labeler Module
    Aliquoter/Labeler Module – Automatic sample aliquoting to secondary tubes using disposable tips. Automatic barcode labeling of secondary tubes.
    TCAutomation Centrifuge Module
    Centrifuge Module – Automatic and continuous centrifugation of samples.
    TCAutomation Decapper Flex Module
    Decapper Flex Module – Automatic cap removal from different types and sizes of sample tubes.
    TCAutomation Recapper Module
    Recapper Module – Automatic capping of different types and sizes of primary and TCAutomation Aliquoter secondary tubes with conical caps.
    TCAutomation Buffer Module
    Buffer Module – Area for samples waiting for validation and for storing empty carriers.
    TCAutomation Cold Storage Module
    Cold Storage Module – Refrigerated storage module from which samples can be automatically retrieved back to the track for re-testing or discharged.
    TCAutomation Perpendicular and Parallel Bypass Modules
    Perpendicular and Parallel Bypass Modules – Analyzer- specific interface modules for point-of-space system connections.
    TCAutomation Robotic Analyzer Interfaces
    Robotic Analyzer Interfaces – Please contact Thermo Fisher Scientific for more information.
    Fully featured solutions for optimized workflow
    Effective sample processing
    Samples are transported within the TCAutomation system in single tube carriers that accommodate various tube types and sizes.
    • Sample tube carriers with an embedded RFID microchip enable faster and more reliable sample identification.
    • Excellent real-time sample tracking possibilities.
    Centralized operation
    TCAutomation controller, based on Windows™ software, offers in a single screen an overview of the entire system.
    • Controller allows users an easy way to monitor large automation configurations.
    • Route definitions are simple to manage and provide flexibility when optimizing the workflow.
    Expandability step by step
    Depending on the floor plan and efficiency requirements, TCAutomation systems can be expanded step by step by adding modules.
    • Expand functionality with additional modules
    • Increase throughput by multiple modules
    • Extend analytical integration by adding analyzer interfaces