Accelerating AML and MDS Treatment Strategies with Rapid NGS in the myeloMATCH Trial
Rapid genomic profiling
In the era of precision oncology, genomic profiling plays a crucial role in the assessment of acute myeloid leukemia (AML) and myelodysplastic syndrome (MDS). However, traditional genomic analysis using next-generation sequencing (NGS) can be time-consuming, taking weeks to provide results and potentially leading to delays in treatment decisions.
Rapid NGS testing addresses this challenge through workflow automation, which enables laboratories to generate a comprehensive molecular profile with next-day results. The data obtained from rapid NGS can inform front-line treatment decisions and facilitate swift patient enrollment into clinical studies.
Video Player is loading.
Current Time 0:00
Duration 1:05
Loaded: 15.15%
Stream Type LIVE
Remaining Time 1:05
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    The NCI Myeloid Assay based on Ion Torrent and Oncomine technology
    Rapid NGS testing has recently been implemented as part of the myeloMATCH trial, which is the National Cancer Institute's (NCI's) precision medicine umbrella trial for AML and MDS patients.
    Through this approach, patients can be assigned to a sub-study within 72 hours of the lab receiving the sample, allowing timely therapy initiation.
    At the heart of the myeloMATCH molecular testing strategy is the NCI Myeloid Assay, run on the Ion Torrent Genexus System. This NGS test enables a comprehensive molecular analysis of DNA mutations and RNA fusions, with results available typically within 72 hours for patient enrollment. With rapid NGS, there is great potential for helping to  improve patient outcomes and expediting the availability of new therapies. 
    Notably, the myeloMATCH program, including the use of Ion Torrent and Oncomine technology for the NCI Myeloid Assay, has received Investigational Device Exemption (IDE) and Investigational New Drug (IND) authorization.
    myelomatch-genexus
    NCI Myeloid Assay performance data
    AMP 2023 Corporate Workshop: Rapid NGS for Genomic Profiling of Myeloid Malignancies
    View this presentation by Cecilia Yeung, MD, Associate Professor, Clinical Research Division, Fred Hutch Cancer Center, to see performance data from the NCI Myeloid Assay with the Genexus System.
    rapid ngs workshop
    ASH 2023 Scientific Poster: Rapid Clinical Mutation Screening for AML Using the Genexus Platform
    See recent performance data for the NCI Myeloid Assay v2, showing high specificity, sensitivity, accuracy, and reproducibility (inter-lab and intra-lab) with sequencing results generated within 48 hours of assay initiation2.
    rapid myeloid ngs poster
    Accelerating treatment strategies
    myeloMATCH is open through the NCI National Clinical Trials Network in the United States and Canada, encompassing over 2,200 sites.
    Cooperative research and development agreements with various pharmaceutical companies will enable access to a range of different drugs to support the myeloMATCH trial.
    By conducting multiple treatment sub-studies tailored to specific genomic types, myeloMATCH has the potential to drive the advancement of exciting new therapies.
    treatment strategies
    Ready to speak to a Thermo Fisher representative?
    We will be happy to answer your questions and provide a demo of our NGS solutions.
    References
    1. Richard F. Little, et al. Umbrella Trial in Myeloid Malignancies: The Myelomatch National Clinical Trials Network Precision Medicine Initiative. Blood (2022) 140 (Supplement 1): 9057–9060.
    2. Shahanawaz Jiwani, et al. Clinical Mutation Screening for AML Using the Genexus Platform. 65th ASH Annual Meeting and Exposition (Poster Presentation) 
    For Research Use Only. Not for use in diagnostic procedures.
    NCI has validated Genexus for investigational use within the MyeloMatch study. Genexus is currently available in the North America for research use only. Not for use in diagnostic procedures.
    1x1 image pixel for data collection