One Lambda HLA PRO, an automated pipetting system for a busy and evolving laboratory can provide a more efficient and flexible option for multiple workflows. Standalone workflows are available that can support NGS library preparation and bead based assays. This system is easily scalable to accommodate variations in sample load while enabling assay precision and consistency. 
A user-friendly interface and walk-away capability liberate laboratory staff from repetitive tasks allowing them to focus on their next priority.
Features
Flexibility - single deck layout for multiple assay automation. Different technologies on one instrument can keep costs and maintenance time low and provide the capability for various workflows.
Accuracy - excellent liquid-level detection and low-volume pipetting for up to 384 samples per run (for bead-based assay automation and 192 sample results per run for NGS sample prep automation).
Easy-To-Use - an intuitive software interface facilitates quick and efficient setup. The touchscreen tool provides visual confirmation for reagent and assay setup at every step of the process.
Let's take a look inside
The modular system offers flexibility by accommodating different workflows on a single instrument, allowing laboratories to automate multiple technologies cost-effectively.
Video Player is loading.
Current Time 0:00
Duration 4:41
Loaded: 0.00%
Stream Type LIVE
Remaining Time 4:41
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    One worktable for all workflows
    Capability of multiple workflows on one platform without any modifications or recalibration to the worktable.
    The system is designed to minimize its footprint in the lab with below-the-deck storage for the centrifuge and waste receptacles. An air-displacement pipettor also eliminates the need for bulky liquid waste containers.
    Worktable
    Touch tool screen
    The touch tool software offers users step-by-step instructions for loading and visual guidance for placing racks, trays, and tips. The racks come fully barcoded for efficient reagent tracking.
    Touch tool screen
    Platform overview
    Plates and tips are loaded in the hotel rack at the back of instrument. Reagent troughs have designated slots and guidance provided with touch screen. Tip and liquid waste stations are designed with efficiency. A shaker and access to the centrifuge round off functionality.
    Platform overview
    Robotic arms
    The capability for simultaneous movement of two arms; Flexible Channel Arm for the tips and Gripper Arm for plate pick up enables faster processes. The different gripper finger designs at the "Exchange reader" provides options for various plate capabilities, providing functionality and flexibility for testing procedures
    Robotic arms
    NGS components
    NGS reagent rack for vials and lids allows efficient storage.
    Magnetic tray for washing steps and filtered tips can be used for NGS assays.
    NGS components
    Thermal cycler
    The automated cycler is included with both bead-based and NGS options.
    Tip box types are switched out for different assays, enabling a quick turnaround for laboratory staff with no additional changes or calibration for another set up.
    Thermal cycler
    Utlizing LABScan3D for bead-based assays
    This configuration enables a LABScan3D to be connected to the HLA PRO, supporting the automation provcess from set up to the reading of the tray. The swivel platform design provides access to LABScan3D for maintenance and servicing.
    HLA PRO and LABScan3D for bead-based assays
    Centrifuge
    The configuration has been designed for efficient use of space in the laboratory – the centrifuge is located bleow the work deck  and protected within a closed cabinet.
    Plates can be lowered down into the centrifuge with a custom counterbalance, to support multiple assays and sample numbers.
    It is easily accessible for cleaning and scheduled maintenance and servicing.
    Centrifuge
    Technical specifications
    Delivering efficiency, flexibility, and performance to meet your laboratory’s evolving needs.
    System overview
    Robotic arms
    Capable of using 2 robotic arms simultaneously
    Deck check
    Utilize a step by step deck loading procedure to help customers load correct labware and reagents, comparing to the expected deck layout and highlight any discrepancies.
    Robotic gripper arm
    The long Z axis on the gripper arm enable the access to below deck; automatic Finger Exchange System with gripper fingers – eccentric and long eccentric
    Door locks
    Doors (hood and centrifuge) lock protect process from free access
    Liquid handling performance (volume range)
    Tip type
    10ul
    50ul
    200ul
    350ul
    1000ul
    Volume precision
    0.5ul,
    10ul
    1.0ul
    50ul
    10.0ul
    200ul
    10ul
    200ul
    10ul
    1000ul
    CV
    ≤6.0%
    ≤1.0
    ≤4.0%
    ≤.3%
    ≤2.0%
    ≤0.2%
    ≤2.0%
    ≤0.5%
    ≤2.0%
    ≤0.3%
    Accuracy
    ±9.5%
    ±2.0%
    ±8.0%
    ±0.5%
    ±2.0%
    ±0.5%
    ±2.0%
    ±0.5%
    ±3.0%
    ±0.0%
    Full configuration
    Dimensions (HxWxD)
    2,301 mm/90.6” x 2,286 mm/90.0” x 785 mm/30.9”
    Weight
    254 kg/535.3 Ibs
    Base configuration
    Dimensions (HxWxD)
    2,301 mm/90.6” x 1,650 mm/65.0" x 785 mm/30.9”
    Weight
    140 kg/306 Ibs
    Service and support
    Our comprehensive service and support facilitate seamless instrument installation, providing you with peace of mind every step of the way. From initial setup to ongoing assistance, our expert team is dedicated to helping you achieve efficient installation and smooth operation of your equipment. With our commitment to customer satisfaction, you can trust us to deliver unparalleled service and support for all your needs.
    Collaborating closely with our global channel partners allows us to offer extensive global support for installation and training, along with comprehensive service plans post-warranty.
    Need help finding the right solution?
    For Laboratory Use.