One Lambda Devyser Accept cfDNA Assay
The One Lambda Devyser Accept cfDNA single-tube NGS assay is a minimally invasive solution that helps provide precise and reliable results, to detect donor-derived cfDNA (dd-cfDNA).
Assay key features and benefits
  • Dual dd-cfDNA distinction: May accurately differentiate the origins of two genetically distinct samples
  • Highly sensitive: Enables the detection of cfDNA at levels as low as 0.1%
  • Robust indel design: Utilizes 50 highly informative and population-independent indel markers. The assay detects and differentiates between two genetically distinct dd-cfDNAs
  • Unified workflow: Simplify your laboratory processes with a streamlined workflow in as little as 45 minutes hands-on time
  • Straightforward data analysis: The user-friendly, dedicated software simplifies data analysis and bioinformatics, and the built-in quality controls facilitate confidence in results
What is the One Lambda Devyser Accept cfDNA Assay?
Minimally invasive and highly robust method for detecting dd-cfDNA
One Lambda Devyser Accept cfDNA is an NGS assay designed to detect and analyze circulating dd-cfDNA in human plasma samples. Its unique indel design allows for accurate results, reduces amplification and sequencing errors. Combined with an efficient workflow, One Lambda Devyser Accept cfDNA is an ideal tool to detect dd-cfDNA and provides early time to result when run in your HLA lab.
On-demand webinar: A Minimally Invasive Test That Can Support Accurately Assigning the Origin of Detected cfDNA
How does the One Lambda Devyser Accept cfDNA Assay work?
The One Lambda Devyser Accept cfDNA assay was developed in conjunction with a nephrologist to deliver efficiency and ease of use with minimal hands-on time allowing for next-day results. Our dedicated software simplifies data analysis and bioinformatics capabilities, allowing for easy sample tracking in a single location. With the ability to import analyzed data into HistoTrac, all information is consolidated, providing clear visibility of trends.
View sample results Fig. 1. Workflow.
Download Advyser Solid Organs Software
From lab technicians seeking efficiency in research to nephrologists gaining insights, Advyser Solid Organs software supports a complete set ecosystem. Utilizing a panel of genetic markers Advyser Solid Organs offers better quantification accuracy, providing new insights in post-transplant research.
Detect dd-cfDNA with ease and condfidence using One Lambda Devyser Accept cfDNA Assay
For Research Use Only. Not for use in diagnostic procedures. Availability of product in each country depends on local regulatory marketing authorization status.