The Ports and URLs related to your instrument's model type are to be entered (whitelisted) on your network firewall by a qualified network engineer. They are not meant to be opened manually from a browser window, doing so will return an error message.
Select the model number of the device you wish to connect. Keep in mind, ALL networking requirements must be met for connectivity to work. If you haven't done so already, please fill out this form to help us better assist you.
Please select model
Ports: 
  • outbound 443
  • outbound 80
Remote Services and Region Settings
File Transfer and User Linking
Note: The * indicates a wildcard.
Ports:
  • outbound 443
  • outbound 80
Remote Services
File Transfer
Firmware Updates
    Remote Services in China
    QS 3/5: Multicast address 224.0.0.251 on port 5353 is used for instrument discovery. Port 7000 outbound calls are used by the QuantStudio Design and Analysis desktop Software to send commands to the instrument.
    Ports: 
    • outbound 443
    • outbound 80
    Remote Services and Region Settings
    File Transfer and User Linking
    Note: The * indicates a wildcard.
    Ports:
    • outbound 443
    • outbound 80
    Remote Services
    File Transfer
    Firmware Updates
      Remote Services in China
      QS 3/5: Multicast address 224.0.0.251 on port 5353 is used for instrument discovery. Port 7000 outbound calls are used by the QuantStudio Design and Analysis desktop Software to send commands to the instrument.
      Share
      Click here to print page