Cryo-EM data collection software solution for single particle analysis
Thermo Scientific Smart EPU Software is an AI-powered, cryo-electron microscopy (cryo-EM) software platform that automates data acquisition, optimizes grid screening, and seamlessly integrates with your hardware for accelerated single particle analysis data workflow. 
Compatible with all Thermo Scientific TEMs, the integrated components like Thermo Scientific Smart EPU Plugins, EPU Multigrid Software, and Embedded CryoSPARC Live Software, work together to support autonomous screening and yield results with high throughput.
1024x576
Software portfolio for SPA data collection
Whether you need end-to-end autonomous screening and real-time quality monitoring or standard automated image acquisition, Thermo Fisher Scientific offers a breadth of solutions to accelerate your single particle analysis data collection.
Smart EPU Software features
Smart technology optimizes decision-making in SPA data collection
Plug into AI-based algorithms, faster screening, and workflow automation with Smart EPU Plugins.
Decision-making can be made easier with:
  • Smart Grid Square Selection for an AI-driven selection and categorization of grid squares 
  • Smart Hole Finder for input-free identification of foil holes, even on challenging grids 
  • Smart Filter for automatic selection of the best quality holes 
  • Smart Ice Thickness Prediction for automated classification of foil holes depending on predicted ice thickness 5. 
  • Smart Focus for auto-focus stabilization using a real-time CTF 
  • Smart Grid Skip for automated grid-square skipping with a real-time contrast transfer function (CTF)
  • Smart Stage Time for stage waiting time optimization
Additionally, advanced users have the opportunity to develop and share their own plugins through an open application programming interface (API). 
Smart EPU Software screenshot showing grid atlas squares used in cryo-EM data collection.
Benefits of screening with Smart EPU Software
Easy adoption 
A user-friendly interface enable fast training and setup, requiring no expertise to identify and exclude bad foil holes.
Maximized efficiency
An autonomous sequence for grid screening uses AI-driven algorithms to reduces microscope time spent assessing sample quality samples.
Workflow optimization
The ability to screen two full cassettes per day accelerates time to results and increases cost savings.
High data quality
Automatically excludes sub-optimal areas for more reliable results.
Resources
Featured resource
Technical specifications for Smart EPU Software
Dive deeper into the innovations behind Smart EPU Software and see how it can enable a more automated experience for cryo-EM single particle analysis data acquisition.
Additional resources
Webinars
What’s new in Smart EPU Software?
This tutorial introduces two new features now available in Smart EPU Software:
  1. A targeting correction routine for the improvement of atlas-tile stitching
  2. A beam-centering sequence option for Thermo Scientific Glacios Cryo-TEMs with FFI, which maintains the beam center without user supervision during EPU session runs 
Additional Smart EPU Software tutorials
What’s new on Smart EPU update: Discovery Viewer
This tutorial demonstrates how to use Discovery Viewer to identify grid areas for high-quality data acquisition.
Video Player is loading.
Current Time 0:00
Duration 2:34
Loaded: 0.00%
Stream Type LIVE
Remaining Time 2:34
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
      1. Now Playing
        Up NextWhat’s new on Smart EPU update: Discovery Viewer
        This tutorial demonstrates how to use Discovery Viewer to identify grid areas for high-quality data acquisition.
      2. Now Playing
        Up NextWhat’s new on Smart EPU 3.10 update: Smart Hole Finder
        This tutorial demonstrates how to use new capabilities in Smart EPU: Optimize Optics, Smart Grid Square Selection for gold grids, Smart Hole Finder, and a measurement tool.
      3. Now Playing
        Up NextWhat’s new in Smart EPU update: CryoSPARC Live™ Multigrid | August 2024
        This tutorial demonstrates the latest features of Smart EPU Software included: integrated data management, new automation parameters and multigrid support for embedded CryoSPARC Live
      4. Now Playing
        Up NextWhat’s new on Smart EPU update: Smart Ice Thickness Prediction
        This tutorial demonstrates how to use three new capabilities in Smart EPU: Plasmon Peak Imaging , Smart Ice Thickness Prediction, and Tilted Acquisition.
      5. Now Playing
        Up NextWhat’s new on Smart EPU update: Smart Selection of Grid Square Plugin with categorization
        This tutorial demonstrates how to use Smart Selection of Grid Square to automatically select and categorize grid squares.
      6. Now Playing
        Up NextHow to get started with Smart EPU Software (EPU 3.6) with Embedded CryoSPARC Live-Nov 2023
        This tutorial demonstrates how to get started with Smart EPU Software (EPU 3.6) with Embedded CryoSPARC Live. This software enables users processes images on the fly to assess sample quality and accelerate structural determination.
      7. Now Playing
        Up NextWhat’s new on Smart EPU update Smart Selection of Grid Squares
        This tutorial demonstrates the latest features of Smart EPU Software including the new Athena selection window, Smart Selection of grid squares, and automatic placement of the autofocus area.
      8. Now Playing
        Up NextHow to get started with Smart EPU Software with Embedded CryoSPARC Live
        This tutorial demonstrates how to get started with Smart EPU Software with Embedded CryoSPARC Live. This software enables users processes images on the fly to assess sample quality and accelerate structural determination.
      9. Now Playing
        Up NextWhat’s new on Smart EPU update: EPU Multigrid Software
        New option for EPU 2.10 and later: EPU Multigrid. EPU Automated Data Acquisition Software for Single Particle Workflow.
      10. Now Playing
        Up NextWhat’s new on Smart EPU update: Smart Filter, Fast Screening and Auto-create sessions
        This tutorial demonstrates how to use Smart Filter, Fast Screening and Auto-create sessions to facilitate sample screening in single particle analysis.
      11. Now Playing
        Up NextWhat’s new on Smart EPU update: Image Shift Calibration
        This tutorial demonstrates the latest features of Smart EPU Software for SPA included: TIFF LZW output for Falcon-4i cameras, guided Image Shift Calibration, and clean up functions.
    Software updates
    Release management
    Thermo Fisher Scientific offers a constant stream of software improvements and updates, ensuring you can achieve your goals as quickly and easily as possible. 
    For Research Use Only. Not for use in diagnostic procedures.