prrsv-diagnostics-showcase-image-230x195
PRRSV diagnostic solutions that evolve with your needs
Veterinarians and producers rely on your guidance to help ensure the health of their pig herds. And their main request is clear: reliable, timely test results from a variety of sample types.
We can partner with your lab to design a solution that adapts to your evolving needs and produces actionable PRRSV (porcine reproductive and respiratory syndrome virus) test results. We develop unique diagnostic schemes for PRRSV, with sampling, sample preparation tools, and workflows for real-time PCR and ELISAs.
The use of both molecular and serological tests gives the most complete picture of herd status because each measures a different aspect of the PRRS virus. 
Sampling
The GenoTube Livestock Swab is designed to quickly dry and stabilize fluid samples, enabling you to ship high-quality samples to the lab
  • Easy transport and shipping—no cooling required
  • Helps reduce costs for cooled transport or time-critical transport requirements, and costs for sample storage
  • Validated for oral fluids and blood samples
  • Optimized sample extraction protocols for PCR and ELISA
  • Recommended by FLI in Germany as sampling tool for ASF
 See the video ›
 Download scientific poster ›
genotube
Oral fluid sampling that is animal-welfare friendly is already standard practice in some countries
  • A single oral fluid sample can be used to test for multiple respiratory pathogens
  • Used as a sample matrix, testing oral fluids allows for the detection of nucleic acid from PRRSV in an early infection stage with the same performance (sensitivity) as blood, serum, and other validated matrices
  • Provides an opportunity to potentially increase the number of pigs tested
  • Option to use GenoTube to collect oral fluid samples from wringed robes
  • Workflow compatible with existing laboratory processes
 View webinar ›
 Download product flyer ›
 Download scientific poster ›
oral-fluid-pig-small
We’ve created a series of best practice guides that outline how, where, and when to take samples from pigs for submission to diagnostic laboratories.
See all guidelines for taking diagnostic samples from pigs
ELISAqPCR
Individual sampleIndividual samples
  • Serum
  • Blood/serum
  • Oral fluid
  • Semen
  • Tissue
  • Organ
  • Swab
  • GenoTube
 Pooled samples 
  • Oral fluid
  • GenoTube
  • Serum
  • Semen
Real-time PCR tests for PRRS virus detection
Real-time PCR is the most sensitive method to provide clear proof of PRRS infection or non-infection. In cases where a positive result is found, the nucleic acid can be used for sequencing. We have access to PRRS virus samples from different parts of the world, enabling us to follow and monitor genetic changes to the virus. Applied Biosystems VetMAX PCR testing detects all currently known North American (NA) and European (EU) strains.
VetMAX PRRSV 3.0 Reagents and Controls
Poster from the 2021 Leman Swine Conference
Due to the highly mutating nature of PRRSV and newly circulating strains, a project was launched to update the currently on-market PRRSV assay with a new molecular design to provide higher sensitivity towards the new variants.
 Download Poster ›
The VetMAX PRRSV EU & NA 2.0 Kit
A second generation of real-time RT-PCR kit to detect and differentiate European and North American genotypes of porcine reproductive and respiratory syndrome virus (PRRSV) in a variety of swine samples
  • Ready to use triplex RT-PCR kits that simultaneously amplify target RNA of EU and NA PRRSV and exogenous internal control
  • Advanced design targets all four subtypes of EU PRRSV circulating strains plus specific detection of NA PRRSV strains, including PRRSV HP strain
  • LDRT-PCR for PRRSV targets has been evaluated at 9 copies/RT-PCR in fast mode, in mono and co-infection
  • Individual: serum, whole blood, tissue, oral fluid & semen. Pools of up to 5: serum, whole blood, and semen.
  • Simple and fast workflow yields results in one hour 50 minutes
  • Evaluated on 780 field samples of 13 origins tested as part of 5 external studies conducted in Austria, Belgium, Slovenia, Spain, and UK
Find out more
 Download product flyer ›
Applied Biosystems VetMAX PRRSV NA and EU reagents are used for the amplification of NA and EU PRRSV nucleic acids
  • Best suited for regions where PRRS NA strain is most prevalent
  • Easy to use and compatible with a wide variety of sample matrices
  • Included Applied Biosystems Xeno RNA internal positive control enables monitoring of nucleic acid isolation efficacity as well as PCR inhibition
  • Extensive bioinformatics capabilities
  • Standardized results to help reduce variability
  • Part of fully integrated, single-vendor workflow
Find out more
 Download flyer ›
Optimized qPCR workflow combining sample preparation and amplification solutions
The Applied Biosystems MagMAX sample preparation system helps make purifying nucleic acids from pigs’ samples convenient, flexible, and efficient. MagMAX nucleic acid isolation kits along with Thermo Scientific KingFisher Express magnetic particle processors assist the diagnostic laboratory in obtaining the best possible PCR result from the sample. Whether it’s an oral fluid sample collected with a GenoTube Livestock swab, a blood sample, or a tissue sample, the goal is to enable a high level of reliability in the results.
For more information, see "Sample Extraction" solutions web page ›
WPR1917-arrow-graphic
Advantages of the PrioCHECK PRRSV Antibody ELISA Kit
The PrioCHECK PRRSV Antibody ELISA Kit shows high correlation with the most frequently used ELISA in the field, and is therefore a powerful alternative in the PRRSV serology test market. Thermo Fisher Scientific offers a comprehensive portfolio of PRRSV solutions to support your lab's unique ELISA workflow requirements.
ELISA test for detection of antibodies against PRRS virus
TheApplied Biosystems PrioCHECK Porcine PRRSV Ab Strip kitis an ELISA test for monitoring and surveillance of PRRSV in pig herds.
  • Designed for the detection of porcine antibodies against both North American and European serotypes of PRRSV in serum samples
  • Plates coated with ORF 7 protein help ensure highly sensitive and specific detection of type 1 and 2 virus serotypes
  • Shows high correlation with the most frequently used ELISA in the field
  • Fast and robust test suited for testing of large numbers of samples
  • Procedure consists of a simple four-step ELISA protocol
  • Conveniently analyze 90 prepared samples in approximately 150 minutes
Available ELISA workflow solutions and instruments include the ELISA washer, ELISA reader, and ELISA interpretation software (SkanIt). Please contact us for more information.
 Download product flyer ›
 Download application note ›
Webinars
Watch this free on-demand webinar and see how your choice of PRRSV test impacts pig herd health & economics. Our presenters explain how to select the correct PRRSV diagnostic tools for your lab, detail the benefits of ELISA screening solutions, and answer questions about best practices, test result interpretation, and more.
 View webinar ›
This webinar will help you answer many practical questions concerning the use of oral fluids and shows the results of two porcine reproductive and respiratory syndrome (PRRS) diagnostic studies in the field using oral fluids. It will also provide you with details regarding the detection of PRRSV in the field via oral fluids, practical tips for oral fluid sampling, and the interpretation of PCR test results for the detection of PRRSV.
 View webinar ›
Ordering information
ELISA
Catalog # Name Size Price (USD) Qty
7610880 PrioCHECK™ Porcine PRRSV Ab Strip Kit Each -
PCR
Catalog # Name Size Price (USD) Qty
A53170 VetMAX™ PRRSV 3.0 Controls Each
162.00
4405548 VetMAX™ PRRSV NA & EU Controls Each
180.00
A52576 VetMAX™ PRRSV 3.0 Reagents Each
1,258.00
Sample Extraction
Sampling
Catalog # Name Size Price (USD) Qty
9062010 GenoTube Livestock Swabs 1 piece
4.17
For more information
To find out more about our animal health products, click the button below.
Contact a product specialist
For Veterinary Use Only. For In Vitro Use Only. Regulatory requirements vary by country; products may not be available in your geographic area.
For Veterinary Use Only. For In Vitro Use Only. Regulatory requirements vary by country; products may not be available in your geographic area.