Search Thermo Fisher Scientific
The Opgal EyeCGas® Multi Optical Gas Imaging (OGI) Camera is a highly sensitive handheld OGI camera. It quickly identifies fugitive gas emissions for over 400 hydrocarbon gases and volatile organic compounds (VOC), including methane and CO2. The EyeCGas Multi OGI Camera is intrinsically safe and complies with the EPA’s OOOOa/b/c regulations.
Certified and classified for hazardous environments, the EyeCGas Multi Optical Gas Imaging Camera is used globally for LDAR compliance. With a wide range of unique features, the EyeCGas Multi OGI Camera helps detect fugitive gas emissions and target gas leaks for repair inside facility limits without the need of a hot permit. The EyeCGas Multi OGI Camera also is capable of video and audio recording and streaming, helping maintain safety during use.
The EyeCGas Multi OGI Camera is designed to deliver remarkable performance and safety for the oil and gas industry. With advanced connectivity, intrinsic safety certifications, and exceptional sensitivity for gas leak detection, this camera sets the standard for optical gas imaging. Its LDAR-ready capabilities, versatile thermographic imaging, and built-in gas quantification help to ensure comprehensive usability and reliability. Explore the advanced features that make EyeCGas Multi OGI Camera a leading choice for environmental monitoring and industrial safety.
Gas leak detection is critical across various industrial settings to ensure safety, prevent environmental damage, and avoid costly fines. Our advanced gas leak detection cameras are designed to meet the rigorous demands of oil refineries, LNG terminals, transmission pipelines, and offshore oil rigs. These cameras provide quick and safe detection and visualization of fugitive gas emissions, enabling prompt repair and maintenance. Explore how our solutions can help you maintain operational integrity and safety in even the most challenging environments.
Service and product availability may vary by country and are subjected to varying regulatory requirements. Please contact your local sales representative for availability.