Search Thermo Fisher Scientific
Use this comparison chart to see which system is right for you.
|
21PlusHD |
Iplus! |
Cast film |
✓ |
✓ |
Co-extruded film |
✓ |
|
Sheet extrusion |
✓ |
✓ |
Biax film |
✓ |
|
Converted products |
✓ |
|
Extrusion coating (total only) |
✓ |
✓ |
Extrusion coating (base/coat/total) |
✓ |
|
Tandem extrusion coating |
✓ |
|
Roll/blade coating |
✓ |
✓ |
Building materials |
✓ |
|
Paper |
✓ |
|
Nonwovens |
✓ |
✓ |
Patch coating |
✓ |
|
Vinyl calendering |
✓ |
✓ |
Roofing |
✓ |
|
Abrasives |
|
|
|
Integra |
Mark III |
C-Frame |
Box Beam |
PROSIS IR Transmission |
|
✓ |
|
|
PROSIS IR Reflection |
|
✓ |
|
|
PROSIS IR Dual Reflection |
|
✓ |
|
|
Beta Plus HE |
✓* |
✓* |
|
|
PM Roofing Sensor |
|
|
✓ |
✓ |
Xray Plus |
✓* |
✓ |
|
|
ShadowMaster |
|
✓ |
|
|
*Available on Iplus! platform |
21PlusHD |
Iplus! |
Range |
Measurement |
Measurement Principle |
|
Beta Plus HE KR85 |
✓ |
✓ |
5–1600 gsm |
Basis weight |
Isotope |
BetaPlus SR90 |
✓ |
✓ |
100–7000 gsm |
Basis weight |
Isotope |
PM Roofing SR90 |
✓ |
|
100–7000 gsm |
Basis weight |
Isotope |
Prosis |
✓ |
|
1–2000 µm depending on material |
Basis weight or direct thickness |
NIR spectroscopy |
ShadowMaster |
✓ |
|
100–6500 µm |
Direct thickness |
Laser |
Xray Plus |
✓ |
|
20–50,000 gsm |
Basis weight |
X-ray |
|
Integra |
Mark III |
C-Frame |
Box Beam |
PROSIS IR Transmission |
|
✓ |
|
|
PROSIS IR Reflection |
|
✓ |
|
|
PROSIS IR Dual Reflection |
|
✓ |
|
|
Beta Plus HE |
✓* |
✓* |
|
|
PM Roofing Sensor |
|
|
✓ |
✓ |
Xray Plus |
✓* |
✓ |
|
|
ShadowMaster |
|
✓ |
|
|
*Available on Iplus! platform |
21Plus! |
Iplus! |
Max Sheet Width |
|
Mark III Industrial Scanner |
✓ |
✓* |
393.7 in. (10m) |
Mark III Hazard Reduced Scanner |
✓ |
|
148 in. (3749mm) |
Mark III Industrial Scanner for ShadowMaster Sensor (includes roll) |
✓ |
|
118 in. (2997mm) |
Mark III Vertical Passline Industrial Scanner—CAN BE TILTED +/- 45 DEGREES FROM VERTICAL |
✓ |
|
148 in. (3749mm) |
Mark III Wide Gap Industrial Scanner |
✓ |
|
148 in. (3749mm) |
Abrasive Scanning C-Frame—ALWAYS VERTICAL |
✓ |
|
80 in. (2032mm) |
Stainless Steel Roofing Scanning (High Moisture Content) C-Frame |
✓ |
|
80 in. (2032mm) |
Integra Scanner |
✓ |
|
88 in. (2236mm) |
Box Beam |
✓ |
|
80 in. (2032mm) |
*Mark III with iPlus! Platform available up to 160 in (4064mm) |
|
Integra |
Mark III |
C-Frame |
Box Beam |
PROSIS IR Transmission |
✓* |
✓ |
|
|
PROSIS IR Microbeam |
|
✓ |
|
|
PROSIS IR Reflection |
|
✓ |
|
|
PROSIS IR Dual Reflection |
|
✓ |
|
|
Beta Plus HE |
✓* |
✓* |
|
|
PM Roofing Sensor |
|
|
✓ |
✓ |
Xray Plus |
|
✓ |
|
|
ShadowMaster |
|
✓ |
|
|
*Available on Iplus! platform |
For Research Use Only. Not for use in diagnostic procedures.