Biotechnology in food production
Biotechnology increasingly extends its reach into very diverse sectors, including pharmaceuticals, fuels, plastics, materials, and food products. Biofood products include novel enzymes that have a profound influence on food texture and flavor, and supermarket shelves now routinely carry alternative proteins like mycoprotein made by fermentation as replacements for traditional meat and dairy. For bio-based foods to become a cost-effective alternative to animal products the industry will need to optimize production efficiency in the manner that the pharmaceutical industry has, and it is here that Process Analytical Technology (PAT) plays a pivotal role. PAT revolves around the real-time monitoring of critical quality attributes (CQAs) and process parameters (CPPs) to elevate production efficiency in the biofood industry.
Biofood products:
  • Enzymes: Traditionally sourced from animal by-products and plant extracts, enzymes have evolved with the advent of microbial fermentation, revolutionizing food attributes.
  • Alternative Proteins: Notably, mycoprotein made through fermentation has disrupted the food industry by offering sustainable dietary alternatives to conventional meat, seafood, and dairy products.
Challenges and solutions:
  • The biofood industry grapples with the dual challenges of addressing consumer concerns and optimizing production efficiency.
  • Process Analytical Technology (PAT): This innovative approach centers on the real-time monitoring of critical quality attributes (CQAs) and process parameters (CPPs) to bolster production efficiency.
Gas analysis with Mass Spectrometry (MS)
  • Gas analysis, particularly through Mass Spectrometry (MS), assumes a pivotal role in the real-time monitoring of fermentation processes.
  • MS delivers exceptional precision in measuring the respiratory gases oxygen, carbon dioxide, inert gases like nitrogen and argon, and a spectrum of volatiles including methanol and ethanol.
  • The integration of MS with a multi-stream sample selector amplifies its efficiency by enabling the rapid monitoring of multiple fermentors by a single analyzer thereby driving down the cost of PAT implementation.
  • Online analytics, complemented by the precision of MS, spearheads a new era of bio-based food innovation, squarely addressing the surging demand for sustainable dietary choices.
  • From the transformative impact of enzymes on food properties to the ascendancy of alternative proteins in reducing reliance on traditional animal-derived products, the biofood industry charts a course toward a brighter and more sustainable future.
Featured products
Prima Pro
Prima PRO
Learn more about gas analysis and biofood production
Thermo Scientific Prima PRO is currently the world’s leading process mass spectrometer, designed to operate reliably in harsh production environments and backed up with an industry-best three year warranty. Features include:
  • Scanning magnetic sector technology for fast, flexible, precise multi-component analysis.
  • Concentrations of respiratory gases, inerts and volatiles can be measured in seconds with high levels of precision.
  • 32- or 64-port Rapid Multistream Sampler (RMS) for fast, reliable monitoring of multiple fermenters and bioreactors.
  • 21 CFR Part 11 compliant GasWorks software.
  • Built-in air conditioner means Prima PRO can operate reliably over a wide ambient temperature range.
Learn more about the Prima PRO
Prima BT
Prima BT
Scalable solution
To support efforts from R&D through to full production scale, we have magnetic sector mass spectrometry solutions in the Thermo Scientific Prima BT Bench Top Process Mass Spectrometer and the Thermo Scientific Prima PRO Process Mass Spectrometer.
Learn more about the Prima BT
Featured Resources
How online process analytics are assisting the biofoods industrys
How online process analytics are assisting the biofoods industrys
Learn more
FIVE REASONS to use Process Mass Spectrometry in Biotechnology
Five Reasons to use Process Mass Spectrometry in Biotechnology
Learn more
Video spotlight
Video Player is loading.
Current Time 0:00
Duration 2:15
Loaded: 7.30%
Stream Type LIVE
Remaining Time 2:15
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Biofoods webinar
    Thermo Fisher Scientific joins forces with Geltor to spotlight the next leap in BioFoods collagen fermentation
    Discover the intersection of sustainability and innovation in "Exploring Collagen Fermentation: The Future of Biofoods." This engaging session unveils the transformative power of biofood technology, promising a future where food production aligns with eco-friendly practices. Dive into a thought-provoking discussion with industry experts Dan Merriman and Andrew Saarni, and see how today's scientific breakthroughs are crafting tomorrow's dining table.
    Join the revolution in sustainable eating. Be part of the change—watch the webinar today.
    Register
    Contact us
    *
    *
    *
    *
    *
    *
    *
    *
     Please confirm you would like to receive marketing and promotional email messages about Thermo Scientific products and services.
    This site is protected by reCAPTCHA and the Google Privacy Policy and Terms of Service apply.
    Style Sheet for Global Design System
    Style Sheet for Komodo Tabs
    Style Sheet for Global Design System