Search Thermo Fisher Scientific
Prior to the development of Process Analytical Technology (PAT), process monitoring systems lacked real-time measurement and relied on manual sampling and feeding, which increased the risk of contamination and batch failures. The Thermo Scientific MarqMetrix All-In-One Process Raman Analyzer with Thermo Scientific MarqMetrix BallProbe Sampling Optics and Thermo Scientific Lykos PAT Software enables users to perform in-line, real-time measurements of Critical Process Parameters (CPPs) and Critical Quality Attributes (CQAs) for continuous manufacturing.
Save time and costs with:
Designed for out-of-the-box use, the MarqMetrix All-In-One Process Raman Analyzer enables highly accurate Raman measurements in less than 15 minutes at the point of need. Open, unpack, make three connections, press the one-button, and go. Factory calibration ensures precise and continuous monitoring on the go. The analyzer can be easily integrated into your existing process monitoring and eliminates the need for costly technical expertise.
The MarqMetrix All-In-One Process Raman Analyzer provides real-time, in-line process monitoring, eliminating the need for sample preparation in markets where time-to-results is critical. With simple, reliable, and precise measurements, the MarqMetrix All-In-One Process Raman Analyzer supplies manufacturers deeper process insights, allowing them to optimize processes with no downtime.
The MarqMetrix All-In-One Process Raman solution offers Fiber Head and BallProbe Sampling Optics that fit any need due to a wide array of easily interchangeable BallProbes. Swapping the BallProbe is easy: simply unscrew the fastener, remove the probe, swap in another, and tighten it down again. Our tapered fitting allows for easy indexing and requires no alignment nor any need to recalibrate the system. The analyzer’s lineup of standard Fiber BallProbes allows measurement of compounds in any form—e.g., solids, liquids, gas, slurries, pastes, or gels. It can measure by contact or immersion with its performance half inch BallProbe and/or standard BallProbes.
Every MarqMetrix All-In-One system leaves our facility factory calibrated, ensuring continuous and precise analysis. Our ongoing calibration study demonstrates the stability and performance of our Raman analyzers without recalibration for over 5 years.
The MarqMetrix All-In-One Process Raman Analyzer has been stable and accurate within ±0.1 cm-1 (±1.5%), 5x better than reported ASTM reference accuracy without recalibration. Its outstanding stability allows you to run continuously with zero downtime with a calibrated process Raman analyzer. Focus on process quality and efficiency, not hardware maintenance.
Lykos PAT Software, a remote control and monitoring software, is designed to simplify the complexities of chemometrics and provide an intuitive workflow to streamline processes. Coupled with Lykos PAT Software – Compliance, users are empowered to retrieve process data like never before and attain cGMP and 21 CFR Part 11 FDA compliance.
For Research Use Only. Not for use in diagnostic procedures.