NIR Process analyzer
Remotely monitor your materials or process using simultaneous fiber optic multiplexing technology and integrated industrial communication for real-time feedback and control with the Thermo Scientific Antaris MX FT-NIR Process Analyzer. The Antaris MX FT-NIR Process Analyzer provides a complete, fit-for-purpose fiber optic interface solution for point-of-use materials analysis and online process monitoring applications.
This easy-to-use NIR analyzer allows a single process to be monitored at multiple points or multiple processes to be monitored simultaneously. The Antaris MX Process Analyzer uses automated background collection to eliminate removing the probe from the process stream which provides continuous uninterrupted analysis of your materials.
Thermo Scientific Antaris MX FT-NIR Process Analyzer
Antaris MX FT-NIR Process Analyzer features
  • 2 or 4 built-in fiber optic channels for simultaneous data collection with no moving parts
  • Continuous automated internal background collection
  • SMA905 industry-standard connectors for fiber optics — choose the probe right for your process
  • OPC, ModBus TCP/IP, 4-20mA and Digital I/O for process communications
  • Recommended for in-process liquid analysis, in-process solid analysis, laboratory liquid analysis, and laboratory solid analysis
  • Thermo Scientific ValPro Instrument Qualification Package
Antaris MX FT-NIR Process Analyzer ordering information
The Antaris MX FT-NIR process analyzer is sold as a configured system designed to meet your specific application needs.