Profile more biomarkers with less sample
Invitrogen ProcartaPlex assays are antibody and magnetic bead-based multiplex immunoassays that leverage Luminex xMAP technology to enable the simultaneous detection and quantitation of up to 80 protein targets in a single well with only 6.3–50 µL sample of plasma, serum, cell culture supernatants, or other bodily fluids. The assays cover more than 600 cytokines, chemokines, growth factors, and other protein targets from a range of species.
With a broad menu of preconfigured panels (2- to 80-plex) and customizable options, ProcartaPlex assays provide high flexibility to serve your individual research needs. Whether investigating inflammation, immunology, cytokine profiling, oncology, or infectious diseases, you can rely on the accuracy, sensitivity, reproducibility, and versatility of ProcartaPlex multiplex assays to gain deeper insights and uncover complex biological processes.
ProcartaPlex multiplex assays provide a high-throughput (96- and 384-well format), cost-effective and time-saving solution for biomarker analysis and disease research.
ProcartaPlex assay formats
Dual Reporter Panels
NEW Explore the first-commercially available ProcartaPlex Dual Reporter assay that enables the simultaneous quantitation of the phosphorylated and total forms of up to 8 signaling proteins on the same bead.
Why ProcartaPlex assays?
Flexibility
Get exactly the panel you want. More than 90% of ProcartaPlex assays can be combined with one another and allow for the creation of highly individual and high-plex panels (≤ 80-plex).
Reproducible results
Each ProcartaPlex assay is manufactured in an ISO 9001 certified facility and undergoes rigorous QC testing to help ensure reproducibility and lot-to-lot consistency.
Scalability
Achieve reproducible performances regardless of plex size and correlate data from differently sized panels and simplex kits.
High specificity and sensitivity
All ProcartaPlex assays are extensively and individually tested for specificity and detection of native endogenous protein as well as for combinability, interference, and cross-reactivity.
Save resources
ProcartaPlex multiplexing assays require smaller sample input and less hands-on time than single-analyte assays, helping reduce time to results, cost and labor.
Free analysis software
ProcartaPlex Analyst App is free software available through Thermo Fisher Connect with 1 TB of cloud storage at no charge.
ProcartaPlex immunoassay data
ProcartaPlex assay performance on Luminex xMAP INTELLIFLEX compared to Luminex 200 and FLEXMAP 3D
(A) Human IL-8 standard curve data shows excellent correlation between INTELLIFLEX and established Luminex instruments. Acquisition on INTELLIFLEX was done on low PMT mode (LPMTLX) to compare with Luminex 200 (LX200) and on high PMT mode (HPMTFM) to compare with FLEXMAP 3D (FM3D). (B) Human IL-8 total protein measurements from various samples (unstimulated and stimulated PBMCs) show excellent correlation and consistency when data was acquired on INTELLIFLEX vs Luminex 200 and FLEXMAP 3D. 
ProcartaPlex immunoassay results
“Our core facility has been using the Invitrogen Th1/Th2 Cytokine 36-Plex Mouse ProcartaPlex Panel on tissue lysates. We have many customers who have been pleasantly surprised by the reproducibility of the assay and some have commented: ‘This assay nicely validated what we have found in the gene expression.'”
―Diane Bender, PhD
The Bursky Center for Human Immunology and Immunotherapy Programs
Washington University School of Medicine
Immunoassay Research Areas
Explore our immunoassays for different research areas, including immuno-oncology, neurobiology, isotyping and immunoglobulin characterization, hallmarks of cancer, CAR-T cells, and more.
Artist rendition of antibody binding to cells Immunoassay Research Areas
Access the largest multiplex panels on the market
Transform your way of biomarker discovery and validation. With the largest ready-to-use panels on the Luminex platform—the ProcartaPlex Human Immune Response Panel 80-Plex and ProcartaPlex Mouse Immune Response Panel 64-Plex—you can accelerate your screening projects.
Understand the effects of cytotoxins in biological systems
Analyze up to 11 biomarkers (including FDA recommended markers) and highly relevant CYP targets for ADME/Tox studies to monitor liver and kidney disease progression, and drug induced hepatotoxicity and nephrotoxicity.
Higher throughput, less sample input
ProcartaPlex 384-well assays offer optimized reagent formulations specifically developed for smaller reaction volumes. With only 6.3µl of sample, get up to 30,720 data points. Our exclusive lyophilized bead kit format combines the shortest time to result with even less hands-on time compared with liquid bead formulation.
“Like magic, science takes practice, but the end results are so satisfying.”
Read the customer story and discover how Martin Pelletier, PhD overcame his challenge to screen for multiplex biomarkers from a small sample volume and to perform large scale tests with high reproducibility in his translational research.
Martin Pelletier, PhD
Researcher, Division of Infectious and Immune Diseases at the CHU de Québec-Laval University Research Center
Associate Professor, Department of Microbiology-Infectious Diseases, and Immunology
Faculty of Medicine, Université Laval, Canada
The ProcartaPlex Analysis App streamlines the analysis of data from ProcartaPlex assays acquired on Luminex instruments. This user-friendly software simplifies the assignment of lot-specific assay information, offering 4PL/5PL curve fit optimization, group-wise statistical and heat map analysis. Additionally, the app allows export of detailed reports including images for presentations and publications.
Snapshot from the ProcartaPlex Analysis App ProcartaPlex Analysis App
For Research Use Only. Not for use in diagnostic procedures.