Decorative image of POROS family of chromatography columns
POROS analytical chromatography columns are used for the quantification and assessment of purity of biomolecules throughout cell culture and purification processes. POROS A, G, AL, EP, OH, and POROS CaptureSelect columns are polymeric packings designed for high-resolution separation, specifically affinity chromatography of antibodies, proteins, polynucleotides, and other biomolecules with user-immobilized ligands. The resins consist of cross-linked poly[styrene-divinylbenzene] (DVB) flow-through particles for rapid mass transport. This base bead is coated with a cross-linked polyhydroxylated polymer (POROS OH), which is then activated with aldehyde (POROS AL) or epoxide (POROS EP) functional groups.
Figure 1. POROS A, G, and CaptureSelect ligands target multiple IgG epitopes. Figure 1 illustrates the binding sites for affinity ligands Protein A, Protein G, and Protein L. Novel POROS ligands provide alternative specificities that enable rapid binding for the quantitation or purification of engineered immunoglobulins.
Note: POROS affinity chromatography columns are shown in orange.
Ordering information
POROS A, G, and ligands are available in convenient prepacked HPLC columns. The column range addresses analytical and preparative requirements for the quantitation and purification of immunoglobulins or Fc-fusion or HSA-fusion proteins.
Affinity resin columns
POROS™ Prepacked Protein A Affinity Column is based on an immobilized recombinant Protein A functional group designed for high-throughput purification of antibodies.
POROS™ Prepacked Protein G Affinity Column is based on an immobilized recombinant Protein G functional group designed for high-throughput purification of antibodies.
Activated affinity
POROS™ Prepacked Immobilized Aldehyde Affinity Column is based on an aldehyde functional group designed for the immobilization of proteins through free amine groups.
POROS™ Prepacked Immobilized Epoxide Affinity Column is based on an epoxide functional group designed for the immobilization of proteins through free amine groups.
Catalog # Name Size Price (USD) Qty
1602906 POROS™ 20 AL Aldehyde Activated Resin Each
4,045.00
POROS CaptureSelect
Chromatography Learning Lab
Check out the new Chromatography Learning Lab from Thermo Fisher Scientific, providing a robust range of webinars, articles, eBooks, infographics, and other digital resources to help you optimize the use of our purification products in your process.
Explore digital resource
Bioprocessing resources
Access a targeted collection of scientific application notes, case studies, posters, white papers and more for bioprocessing:
For Research Use Only. Not for use in diagnostic procedures.