Designed for ease of use and versatility
The Thermo Scientific HyPerforma Glass Bioreactors offer easy operation and rapid assembly for your research and process development needs. These benchtop glass bioreactors are developed to ensure sterile processes and minimize lost batches.
Glass bioreactor performance
  • Impellers provide maximum mixing with minimum shear force, resulting in a higher average kLa and better cell growth
  • Control can be bidirectional, which allows a single-wall vessel to be used for mammalian applications and fermentation processes (requires additional components)
  • The motor adapter uses coupling windows and an alignment marker for easy assembly
  • Ergonomic head plate design provides easy assembly and disassembly of components for rapid reconfiguration
Glass bioreactor family of products in multiple sizes
Key benefits of glass bioreactor
Glass bioreactors lined up in ascending size, from 1 to 15 L
  • Available in 1, 3, 7, and 15 L sizes
  • Minimal downtime for cleaning
  • Applications include suspension and adherent cell culture processes
  • Appropriate for PD scale to full cGMP manufacturing workflows
Glass bioreactor automation and software
TruBio software uses the advanced DeltaV Live user interface package by Emerson for fully scalable control and is designed to evolve with your process needs, to help you adapt to future advancements.
TruBio software for use with glass bioreactors
The Thermo Scientific HyPerforma G3 family of bioprocess controllers provides flexible and reliable advanced control capabilities. The controllers are fully self-contained, movable units that can be operated alone (for one vessel) or networked for multiple vessels. They are engineered to optimize capital cost for use in non-GMP and CGMP-certified production facilities to production.
Glass bioreactor literature
Data sheet: HyPerforma Glass Bioreactors offer easy operation, rapid assembly, and are manufactured with the highest standards for materials and surface finish
Brochure: Flexible, reliable, and configurable bioprocessing solutions for process development and production operations                                                                   
Application note: Comparative study Part 1: Intuitive bioprocess scale-up from bench scale to pilot scale                                                                                                           
Additional bioprocessing resources
As a trusted partner, we are committed to supporting your journey toward innovative biotech solutions.
Accelerate your therapeutic monoclonal antibody development and manufacturing program.                             
Learn, evaluate, connect.   
Connect with a Bioprocessing representative or request more info.                                                               
For research use or further manufacturing. Not for diagnostic use or direct administration into humans or animals.