Combine exceptional control and measurement to enhance your process
The Thermo Scientific HyPerforma Rocker Bioreactor is controlled by the HyPerforma G3 Lab Controller and TruBio software, providing a complete, ready-to-use solution. The Rocker Bioreactor uses Thermo Scientific Rocker BioProcess Containers (BPCs) which provide up to 25 L of working volume and integrates to the pH+dO2 sensor for combined pH, DO, and temperature monitoring.
Key features of rocker bioreactor
  • Research, process development and seed train production applications
  • Compatible with most cell culture applications
  • Rocking motion is customizable to your specific workflow
  • Optional tray adapter allows the use of 10 L and 20 L BPCs
  • cGMP-compliant capabilities
  • Load cell for weight control
  • pH+dO2 sensor provides measurement and control of critical process parameters: pH, DO, and temperature
HyPerforma rocker single-use bioreactor bioprocess consumables bag
Rocker bioreactor bioprocess containers (BPC)
Designed to create optimal culture conditions
The HyPerforma Rocker bioreactor bag is designed to create optimal culture conditions for rocked cell cultures with multiple ports, allowing an optimized supply of gases, media, and buffers. These bags are manufactured with our proprietary Aegis5-14 film and produced in a cGMP facility.
The proven benefits of Aegis5-14 best support cell culture process development and scale-up needs with confidence and quality, while also providing film consistency through the seed train and production vessels. Rocker bags are available with a sensor puck that includes single-use DO and pH sensors.
Rocker Bioreactor bioprocess containers (BPC)
  • Rocker bag available in 10, 20, and 50 L
  • Leverages pH and DO sensor technology in a compact assembly
  • Puck placement allows the sensor to be welded into the center of the bag, with the reader attached after sterilization
  • Puck sensor technology allows direct measurement comparisons
  • RTD sensor for temperature monitoring
  • Aegis5-14 film provides improved extractables and leachable profiles
Interested in sustainability in bioprocessing? Enjoy the same robust film but now with carbon reduction. Learn more about Thermo Scientific Sustainable film.
Rocker bioreactor automation and controllers
HyPerforma rocker single-use bioreactor with G3Lab Controller and TruBio Software
The Thermo Scientific HyPerforma single-use bioreactor has a quick, simple setup with G3Lab Controller and TruBio Software.
HyPerforma Rocker single-use bioreactor literature
Data sheet: HyPerforma Rocker Bioreactor
Additional bioprocessing resources
As a trusted partner, we are committed to supporting your journey toward innovative biotech solutions.
Accelerate your therapeutic monoclonal antibody development and manufacturing program.                             
Learn, evaluate, connect.   
Connect with a Bioprocessing representative or request more info.                                                               
For research use or further manufacturing. Not for diagnostic use or direct administration into humans or animals.
1x1 image pixel for data collection